Skip to Content
Merck
All Photos(1)

Documents

W269506

Sigma-Aldrich

2-Methylbutyric acid

≥98%, FG

Synonym(s):

NSC 7304

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5CH(CH3)CO2H
CAS Number:
Molecular Weight:
102.13
FEMA Number:
2695
EC Number:
Council of Europe no.:
2002
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
8.046
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 172.515

vapor pressure

0.5 mmHg ( 20 °C)

Assay

≥98%

refractive index

n20/D 1.405 (lit.)

bp

176-177 °C (lit.)

density

0.936 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

cheesy; acidic; fruity

SMILES string

CCC(C)C(O)=O

InChI

1S/C5H10O2/c1-3-4(2)5(6)7/h4H,3H2,1-2H3,(H,6,7)

InChI key

WLAMNBDJUVNPJU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Detection of Maize Mold Based on a Nanocomposite Colorimetric Sensor Array under Different Substrates.: This study focuses on the application of (±)-2-Methylbutyric acid in the development of nanocomposite colorimetric sensor arrays, facilitating the rapid detection of mold in maize. This advancement highlights the compound′s potential in agricultural quality control and food safety (Lin et al., 2024).

  • Regulation mechanism and bioactivity characteristic of surfactin homologues with C14 and C15 fatty acid chains.: Research elaborates on the bioactive characteristics of surfactin homologues involving (±)-2-Methylbutyric acid, underscoring its crucial role in enhancing the microbial production of bioactive compounds. This has significant implications for pharmaceuticals and bioengineering (Su et al., 2024).

  • Multifaceted analysis of the effects of roasting conditions on the flavor of fragrant Camellia oleifera Abel. seed oil.: This paper explores how (±)-2-Methylbutyric acid influences the flavor profile of Camellia oleifera seed oil under various roasting conditions, emphasizing its importance in enhancing food flavors and consumer products (Han et al., 2024).

Biochem/physiol Actions

Taste at 10 ppm

Other Notes

Natural occurrence: Apple, apricot, cider, cocoa, cranberry, grape, grape brandy, oriental tobacco, strawberry, vinegar and whiskey.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

170.6 °F - closed cup

Flash Point(C)

77 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kei M Igarashi et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 32(23), 7970-7985 (2012-06-08)
Odor signals are conveyed from the olfactory bulb to the olfactory cortex (OC) by mitral cells (MCs) and tufted cells (TCs). However, whether and how the two types of projection neuron differ in function and axonal connectivity is still poorly
Vineet K Singh et al.
Applied and environmental microbiology, 74(19), 5882-5890 (2008-08-12)
Staphylococcus aureus is a major community and nosocomial pathogen. Its ability to withstand multiple stress conditions and quickly develop resistance to antibiotics complicates the control of staphylococcal infections. Adaptation to lower temperatures is a key for the survival of bacterial
Ilkeun Lee et al.
Journal of the American Chemical Society, 128(27), 8890-8898 (2006-07-06)
The adsorption and thermal chemistry of (S)-(+)-2-methylbutanoic acid ((S)-2MBA) on Pt(111) single-crystal surfaces was characterized by using temperature programmed desorption (TPD) and reflection-adsorption infrared (RAIRS) spectroscopies. Particular emphasis was placed on the characterization of the chiral superstructures formed upon the
G Hayashida-Soiza et al.
Journal of applied microbiology, 105(5), 1672-1677 (2008-10-03)
To purify and characterize compounds with antimicrobial activity from Pseudoalteromonas haloplanktis inhibition (INH) strain. The P. haloplanktis isolated from a scallop hatchery was used to analyse antibacterial activities. Crude extracts were obtained with ethyl acetate of the cultured broth, after
B Schatowitz et al.
Journal of chromatography, 425(2), 257-268 (1988-03-18)
A method for the simultaneous determination of non-esterified short-, medium- and long-chain fatty acids and other types of metabolically relevant carboxylic acids such as hydroxy, keto, aromatic and dicarboxylic acids in biological material by capillary gas chromatography of benzyl ester

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service