Merck
  • Simultaneous determination of C2-C22 non-esterified fatty acids and other metabolically relevant carboxylic acids in biological material by gas chromatography of their benzyl esters.
Simultaneous determination of C2-C22 non-esterified fatty acids and other metabolically relevant carboxylic acids in biological material by gas chromatography of their benzyl esters.
Journal of chromatography (1988-03-18)
B Schatowitz, G Gercken
ABSTRACT
A method for the simultaneous determination of non-esterified short-, medium- and long-chain fatty acids and other types of metabolically relevant carboxylic acids such as hydroxy, keto, aromatic and dicarboxylic acids in biological material by capillary gas chromatography of benzyl ester derivatives is described. Sample preparation avoiding incomplete isolation of carboxylic acids consisted of deproteinization and extraction with ethanol, fixation of carboxylic acids as carboxylates, removal of interfering compounds such as neutral lipids by hexane extraction and amino acids, acyl carnitines and other cations by cation-exchange chromatography, derivatization of keto groups of ketocarboxylic acids into O-methyl oximes and benzyl ester formation by reaction of the potassium carboxylates with benzyl bromide via crown ether catalysis. The sample preparation conditions were investigated, showing the usefulness of this method for quantitative determinations. Chromatograms obtained from human serum, human urine and rat heart ventricle and concentrations of carboxylic acids in these specimens are presented.
Sigma-Aldrich
(±)-2-Methylbutyric acid, natural, ≥98%, FG
Sigma-Aldrich
2-Methylbutyric acid, 98%
Sigma-Aldrich
2-Methylbutyric acid, ≥98%, FG