Skip to Content
Merck
All Photos(2)

Key Documents

I7004

Sigma-Aldrich

2-Iodoaniline

98%

Synonym(s):

1-Amino-2-iodobenzene, 2-Aminoiodobenzene, 2-Aminophenyl iodide, 2-Iodobenzenamine, 2-Iodophenylamine, o-Aminoiodobenzene, o-Iodoaniline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
IC6H4NH2
CAS Number:
Molecular Weight:
219.02
Beilstein:
2204899
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

55-58 °C (lit.)

SMILES string

Nc1ccccc1I

InChI

1S/C6H6IN/c7-5-3-1-2-4-6(5)8/h1-4H,8H2

InChI key

UBPDKIDWEADHPP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

2-Iodoaniline is an aromatic amine that can be carbonylated to produce pentacyclic compounds.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Palladium-catalysed carbonylation of 4-substituted 2-iodoaniline derivatives: carbonylative cyclisation and aminocarbonylation
Acs P, et al.
Tetrahedron, 62, 12051-12056 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service