Skip to Content
Merck
All Photos(2)

Key Documents

516139

Sigma-Aldrich

4-Chloro-2-iodoaniline

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H3(I)NH2
CAS Number:
Molecular Weight:
253.47
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

mp

39-43 °C (lit.)

functional group

chloro
iodo

SMILES string

Nc1ccc(Cl)cc1I

InChI

1S/C6H5ClIN/c7-4-1-2-6(9)5(8)3-4/h1-3H,9H2

InChI key

FLEJOBRWKBPUOX-UHFFFAOYSA-N

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

>228.2 °F - closed cup

Flash Point(C)

> 109 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quinazolines. VI. Synthesis of 2,4-diaminoquinazolines from anthranilonitriles.
A Rosowsky et al.
Journal of medicinal chemistry, 13(5), 882-886 (1970-09-01)
Regioselective iodination of aryl amines using 1, 4-dibenzyl-1, 4-diazoniabicyclo [2.2. 2] octane dichloroiodate in solution and under solvent-free conditions.
Alikarami M, et al.
Bulletin of the Chemical Society of Ethiopia, 29(1), 157-162 (2015)
Wing S Cheung et al.
The Journal of organic chemistry, 70(9), 3741-3744 (2005-04-23)
[reaction: see text] An efficient and versatile method for stereoselective synthesis of (E)-3,3-(diarylmethylene)indolinones by a palladium-catalyzed tandem Heck-carbocyclization/Suzuki-coupling sequence is presented. Factors influencing yield and selectivity, namely catalyst, coordinating ligand, and solvent, are detailed.
Stereoselective Synthesis of (E)-3-(Methoxycarbonyl) methylene-1, 3-dihydroindol-2-ones by Palladium-Catalyzed Oxidative Carbonylation of 2-Ethynylanilines.
Gabriele B, et al.
European Journal of Organic Chemistry, 24, 4607-4613 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service