Skip to Content
Merck
All Photos(1)

Key Documents

928283

Sigma-Aldrich

RuPhos Pd G3 ChemBeads

Synonym(s):

Adam’s (2-Dicyclohexylphosphino-2′,6′-diisopropoxy-1,1′-biphenyl)[2-(2′-amino-1,1′-biphenyl)]palladium(II) methanesulfonate, RuPhos-G3-Palladacycle, RuPhos-Pd-G3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C43H56NO5PPdS
CAS Number:
Molecular Weight:
836.37
UNSPSC Code:
12352101
NACRES:
NA.82

form

solid

Quality Level

reaction suitability

reagent type: catalyst
reaction type: Cross Couplings

extent of labeling

~5 wt. % loading of catalyst

SMILES string

NC1=C(C2=C([Pd](OS(=O)(C)=O)P(C3CCCCC3)(C4CCCCC4)C5=CC=CC=C5C6=C(C=CC=C6OC(C)C)OC(C)C)C=CC=C2)C=CC=C1

InChI key

OEVHKTCNQBOMNB-UHFFFAOYSA-N

General description

RuPhos Pd G3 is a third generation (G3) Buchwald precatalyst that can be used in cross-coupling reactions for the formation of C-C, C–N, C–O, C–F, C–CF3, and C–S bonds. It is air-, moisture-, and thermally-stable and is highly soluble in a wide range of common organic solvents. Some of its unique features include lower catalyst loadings, shorter reaction time, efficient formation of the active catalytic species and accurate control of ligand: palladium ratio.

For larger scale uses, product also available in powdered form (763403)

Application

RuPhos Pd G3 can be used as a pre-catalyst in the following protocols:

Palladium-catalyzed Suzuki coupling of 5-p-toluenesulfonyltetrazoles with arylboronic acids to synthesize 1,5-disubstituted tetrazoles.Suzuki-Miyaura catalyst-transfer polycondensation (SCTP) of 3-alkylthiophenes in the presence of N-methylimidodiacetic (MIDA)-boronate monomers. Suzuki-Miyaura-cross-coupling of aminothiophenes.

ChemBeads are chemical coated glass beads. ChemBeads offer improved flowability and chemical uniformity perfect for automated solid dispensing and high-throughput experimentation. The method of creating ChemBeads uses no other chemicals or surfactants allowing the user to accurately dispense sub-milligram amounts of chemical.

Learn more about ChemBeads products

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B Inhalation

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

ChemBeads, catalyst-coated glass beads, dispense solid chemical reagents for nanomole-scale high-throughput reaction screening.

ChemBeads, catalyst-coated glass beads, dispense solid chemical reagents for nanomole-scale high-throughput reaction screening.

ChemBeads, catalyst-coated glass beads, dispense solid chemical reagents for nanomole-scale high-throughput reaction screening.

ChemBeads, catalyst-coated glass beads, dispense solid chemical reagents for nanomole-scale high-throughput reaction screening.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service