Skip to Content
Merck
All Photos(8)

Documents

328774

Sigma-Aldrich

Tris(dibenzylideneacetone)dipalladium(0)

97%

Synonym(s):

Pd2dba3, Pd2(dba)3

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5CH=CHCOCH=CHC6H5)3Pd2
CAS Number:
Molecular Weight:
915.72
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Cross Couplings
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

mp

152-155 °C (lit.)

SMILES string

[Pd].[Pd].O=C(\C=C\c1ccccc1)/C=C/c2ccccc2.O=C(\C=C\c3ccccc3)/C=C/c4ccccc4.O=C(\C=C\c5ccccc5)/C=C/c6ccccc6

InChI

1S/3C17H14O.2Pd/c3*18-17(13-11-15-7-3-1-4-8-15)14-12-16-9-5-2-6-10-16;;/h3*1-14H;;/b3*13-11+,14-12+;;

InChI key

CYPYTURSJDMMMP-WVCUSYJESA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tris(dibenzylideneacetone)dipalladium(0) (Pd2(dba)3) is a versatile compound widely used as a Pd(0) source in various Pd-mediated transformations. Pd2(dba)3 is known for its high reactivity and ability to facilitate oxidative addition reactions. It is prepared by reacting palladium salts with dibenzylideneacetone ligands. It acts as a catalyst for several reactions including Suzuki cross-coupling, Heck coupling, arylation, Buchwald-Hartwig amination, and fluorination. It is often used in catalytic amounts and has been shown to be effective in promoting coupling reactions between aryl halides and boronic acids.

For small scale and high throughput uses, product is also available as ChemBeads (919772)

Application

    Application Guide for Palladium Catalyzed Cross-Coupling Reactions

    Reactant involved in:

    • Synthesis of azepanes
    • Synthesis of nanosized palladium phosphides upon interaction with white phosphorous
    • Preparation of palladium triphenylphosphine carbonyl cluster complexes
    • Precursor for synthesis of functionalized multiwalled carbon nanotube-palladium complexes used as catalysts for Heck coupling reactions
    • Selective carbon-sulfur bond formation via addition of S-S and S-H bonds to alkynes


    Catalyst for:

    • Suzuki cross-coupling reactions
    • PCN- and PCS-pincer palladium complex catalyzed tandem allylation
    • Catalyst for Suzuki coupling of aryl chlorides (eq. 1)
    • Catalyst for Heck coupling of aryl chlorides (eq. 2)
    • Catalyst for arylation of ketones (eq. 3)
    • Catalyst for Buchwald-Hartwig amination of aryl halides (eq. 4)
    • Catalyst for fluorination of allylic chlorides (eq. 5)
    • Catalyst for β-arylation of carboxylic esters (eq. 6)
    • Catalyst for carbonylation of 1,1-dichloro-1-alkenes (eq. 7)
    • Catalyst for conversion of aryl and vinyl triflates to aryl and vinyl halides (eq. 8)
    • Pd source for enantioselective Tsuji Allylations

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Satoshi Suetsugu et al.
Organic letters, 16(3), 996-999 (2014-01-28)
The total synthesis of (-)-aurantioclavine (1) was accomplished based on an intramolecular asymmetric amination of allyl carbonate 3 containing a p-tosylamide group. The reaction using tris(dibenzylideneacetone)dipalladium(0), tBu-phosphinooxazoline, and Bu4NCl in CH2Cl2 gave azepane 2 in 77% yield with 95% enantiomeric
Martin Arthuis et al.
Chemical communications (Cambridge, England), 46(41), 7810-7812 (2010-09-21)
A novel and fully chemo- and stereoselective three component strategy leading to Z-α-chloroacrylates by a Pd(0)-catalyzed reaction of CO (1 atm) with 1,1-dichloro-1-alkenes and various alcohols is disclosed. This catalytic approach compares favourably with the Wittig type strategies as α-chloroacrylates
Matthew H Katcher et al.
Journal of the American Chemical Society, 132(49), 17402-17404 (2010-11-23)
The enantioselective fluorination of readily available cyclic allylic chlorides with AgF has been accomplished using a Pd(0) catalyst and Trost bisphosphine ligand. The reactions proceed with unprecedented ease of operation for Pd-mediated nucleophilic fluorination, allowing access to highly enantioenriched cyclic
The Enantioselective Tsuji Allylation
Douglas C. Behenna and Brian M. Stoltz
Journal of the American Chemical Society, 126(46), 15044?45-15044?45 (2004)
Kawatsura, M.; Hartwig, J. F.
Journal of the American Chemical Society, 121, 1473-1473 (1999)

Articles

JosiPhos CyPF-tBu and palladium give catalyst for alkoxylation of activated heteroaryl halides with primary, secondary, and tertiary alcohols

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service