Skip to Content
Merck
All Photos(1)

Key Documents

924377

Sigma-Aldrich

Palladium(II) Acetate ChemBeads

Synonym(s):

Acetic acid palladium salt, Bis(acetato)palladium, Diacetatopalladium, Diacetoxypalladium

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Pd(OCOCH3)2
CAS Number:
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.82

form

solid

Quality Level

composition

~ 4 wt.% loading of catalyst

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction

core: palladium
reaction type: Heck Reaction

core: palladium
reaction type: Hiyama Coupling

core: palladium
reaction type: Negishi Coupling

core: palladium
reaction type: Sonogashira Coupling

core: palladium
reaction type: Stille Coupling

core: palladium
reaction type: Suzuki-Miyaura Coupling

reaction type: Cross Couplings
reagent type: catalyst

SMILES string

[Pd](OC(=O)C)OC(=O)C

InChI

1S/2C2H4O2.Pd/c2*1-2(3)4;/h2*1H3,(H,3,4);/q;;+2/p-2

InChI key

YJVFFLUZDVXJQI-UHFFFAOYSA-L

General description

Palladium(II) acetate is a homogenous oxidation catalyst. It participates in the activation of alkenic and aromatic compounds towards oxidative inter- and intramolecular nucleophilic reactions. Crystals of palladium(II) acetate have a trimeric structure, having symmetry D3h. Each of the palladium atoms in the crystals are joined to the other two by double acetate bridges. Microencapsulation of palladium(II) acetate in polyurea affords polyurea-encapsulated palladium(II) acetate. It is a versatile heterogeneous catalyst for various phosphine-free cross-coupling reactions. It participates as catalyst in the Heck coupling reaction of pthalides with different alkenes.

Application

Catalyst for an intramolecular coupling of aryl bromides with alcohols giving 1,3-oxazepines. Preparation of cyclic ureas via palladium-catalyzed intramolecular cyclization.

ChemBeads are chemical coated glass beads. ChemBeads offer improved flowability and chemical uniformity perfect for automated solid dispensing and high-throughput experimentation. The method of creating ChemBeads uses no other chemicals or surfactants allowing the user to accurately dispense sub-milligram amounts of chemical.

Learn more about ChemBeads products

For larger scale uses, product also available in powdered form (205869) & (520764)

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Sens. 1A

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Novel formation of 1,3-oxazepine heterocycles via palladium-catalyzed intramolecular coupling reaction.
Ma Chen, et al.
Tetrahedron, 62, 9002-9009 (2006)
Palladium-catalyzed β-arylation of modified vinyl ethers with aryl triflates..
Badone D, et al.
Tetrahedron Letters, 34, 3603-3606 (1993)
Palladium-Catalyzed Amination of Aryl Triflates.
Wolfe J P, et al.
The Journal of Organic Chemistry, 62, 1264-1267 (1997)
The First Stereoselective Palladium-Catalyzed Cyclocarbonylation of β,γ-Substituted Allylic Alcohols.
Brunner M, et al.
The Journal of Organic Chemistry, 62, 7565-7568 (1997)
Versatile Methods to Dispense Sub-Milligram Quantities of Solids using Chemical Coated Beads for High-Throughput Experimentation.
Martin C M, et al.
Organic Process Research & Development, 23(9), 1900-1907 (2019)

Articles

ChemBeads, catalyst-coated glass beads, dispense solid chemical reagents for nanomole-scale high-throughput reaction screening.

ChemBeads, catalyst-coated glass beads, dispense solid chemical reagents for nanomole-scale high-throughput reaction screening.

ChemBeads, catalyst-coated glass beads, dispense solid chemical reagents for nanomole-scale high-throughput reaction screening.

ChemBeads, catalyst-coated glass beads, dispense solid chemical reagents for nanomole-scale high-throughput reaction screening.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service