Skip to Content
Merck
All Photos(3)

Key Documents

481084

Sigma-Aldrich

rac-BINAP

97%

Synonym(s):

(±)-2,2′-Bis(diphenylphosphino)-1,1′-binaphthalene, 2,2′-Bis(diphenylphosphino)-1,1′-binaphthalene, (±)-BINAP, [1,1′-Binaphthalene]-2,2′-diylbis[diphenylphosphine]

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(C6H5)2PC10H6-]2
CAS Number:
Molecular Weight:
622.67
Beilstein:
5321443
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

reaction suitability

reaction type: Cross Couplings
reagent type: ligand
reaction type: Acylations

reagent type: ligand
reaction type: Arylations

reagent type: ligand
reaction type: Buchwald-Hartwig Cross Coupling Reaction

reagent type: ligand
reaction type: C-C Bond Formation

reagent type: ligand
reaction type: Decarboxylations

reagent type: ligand
reaction type: Stille Coupling

mp

283-286 °C (lit.)

functional group

phosphine

SMILES string

P(c8ccccc8)(c7ccccc7)c1c(c6c(cc1)cccc6)c2c3c(ccc2P(c5ccccc5)c4ccccc4)cccc3

InChI

1S/C44H32P2/c1-5-19-35(20-6-1)45(36-21-7-2-8-22-36)41-31-29-33-17-13-15-27-39(33)43(41)44-40-28-16-14-18-34(40)30-32-42(44)46(37-23-9-3-10-24-37)38-25-11-4-12-26-38/h1-32H

InChI key

MUALRAIOVNYAIW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Racemic version of BINAP.

Application

Ligand employed in a palladium-catalyzed arylamine coupling in the preparation of demethylthiocholchines. Used with Cu (II) to catalyze addition of arylsulfonamides to styrenes and olefins. Amination of terpyridines with Pd catalysis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Advanced Synthesis & Catalysis, 345, 15-32 (2003)
Tetrahedron Letters, 47, 5079-5079 (2006)
Akira Sakakura et al.
Nature protocols, 2(7), 1746-1751 (2007-07-21)
A protocol for ester condensation between equimolar amounts of carboxylic acids and alcohols catalyzed by bulky diarylammonium pentafluorobenzenesulfonate is described. We also present procedures for the synthesis of N-(2,6-diisopropylphenyl)-N-mesitylammonium pentafluorobenzenesulfonate. The present ester condensation proceeds well under mild conditions even
Jason G Taylor et al.
Organic letters, 8(16), 3561-3564 (2006-07-28)
[reaction: see text] Regioselective additions of arylsulfonamides to vinylarenes, norbornene, and cyclohexadiene were achieved using a copper-diphosphine catayst under mild reaction conditions. These processes appear to be ligand-accelerated.
Organometallics, 23, 3398-3416 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service