Skip to Content
Merck
All Photos(3)

Key Documents

759546

Sigma-Aldrich

Lithium iron(II) phosphate

greener alternative

powder, <5 μm particle size (BET), >97% (XRF)

Synonym(s):

Ferrous lithium phosphate, Iron lithium phosphate, LFP, Triphylite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiFePO4
CAS Number:
Molecular Weight:
157.76
MDL number:
UNSPSC Code:
26111700
NACRES:
NA.23

grade

battery grade

Quality Level

Assay

>97% (XRF)

form

powder

mol wt

Mw 157.76 g/mol

composition

LiFePO4

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

particle size

<5 μm (BET)

mp

>300 °C (lit.)

density

3.6 g/cm3 (lit.)

application(s)

battery manufacturing

greener alternative category

SMILES string

[Fe+2].[Li+].[P](=O)([O-])([O-])[O-]

InChI

1S/Fe.Li.H3O4P/c;;1-5(2,3)4/h;;(H3,1,2,3,4)/q+2;+1;/p-3

InChI key

GELKBWJHTRAYNV-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

General description

Lithium iron(II) phosphate (LiFePO4), also called LFP, is a crystalline, grey-to-black powder with an olivine crystal structure. This unique structure facilitates efficient diffusion of lithium-ions, thanks to the three-dimensional interconnected iron phosphate tetrahedra that form pathways for lithium ions to diffuse. Additionally, the structure is highly stable, not deforming, even after thousands of cycles of lithium intercalation. Our LiFePO4 powder is manufactured to meet high purity with a tightly controlled particle size of less than 5 μm.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Click here for more details.

Application

LiFePO4 is a widely used cathode for rechargeable Li batteries. It has high energy density, wide operating temperature, thermal stability, long cycle life and application life, environmental friendliness, and safety that make it a promising electrode material for greener and safer lithium-ion batteries.

Features and Benefits

LiFePO4-based batteries can last longer, operate safely and reliably, and power devices in a wide range of temperatures.
✔ Achieves 1000s of cycles
✔ Safer than cobalt or manganese-based cathode materials
✔ Reliable performance

Legal Information

Product of Engi-Mat Co.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Low temperature hydrothermal synthesis of battery grade lithium iron phosphate
Benedek P, et al.
Royal Society of Chemistry Advances, 7(29), 17763-17767 (2017)
Electrochemical noise measurement of a lithium iron (II) phosphate (LiFePO4) rechargeable battery
Astafev E, et al.
Instrumentation Science & Technology, 48(1), 75-85 (2020)
Byoungwoo Kang et al.
Nature, 458(7235), 190-193 (2009-03-13)
The storage of electrical energy at high charge and discharge rate is an important technology in today's society, and can enable hybrid and plug-in hybrid electric vehicles and provide back-up for wind and solar energy. It is typically believed that
Kim, G. T.; et al.
Journal of Power Sources, 196, 2187-2187 (2011)
Enhanced electrochemical performance of lithium iron (II) phosphate modified cooperatively via chemically reduced graphene oxide and polyaniline
Shen W, et al.
Electrochimica Acta, 173, 310-315 (2015)

Articles

Professor Qiao's review explores stable microstructures for lithium metal fluoride batteries, advancing energy storage technologies.

Professor Qiao's review explores stable microstructures for lithium metal fluoride batteries, advancing energy storage technologies.

Professor Qiao's review explores stable microstructures for lithium metal fluoride batteries, advancing energy storage technologies.

Professor Qiao's review explores stable microstructures for lithium metal fluoride batteries, advancing energy storage technologies.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service