Skip to Content
Merck
All Photos(1)

Key Documents

436011

Sigma-Aldrich

Iron(III) phosphate dihydrate

Fe 29 %

Synonym(s):

Ferric phosphate dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FePO4 · 2H2O
CAS Number:
Molecular Weight:
186.85
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

form

powder

Quality Level

composition

Fe, 29%

reaction suitability

core: iron
reagent type: ligand
reaction type: Cross Couplings

functional group

amine
phosphine

SMILES string

O.O.[Fe+3].[O-]P([O-])([O-])=O

InChI

1S/Fe.H3O4P.2H2O/c;1-5(2,3)4;;/h;(H3,1,2,3,4);2*1H2/q+3;;;/p-3

InChI key

BMTOKWDUYJKSCN-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application

Iron(III) phosphate dihydrate (FePO4 x 2H2O) can be used as a catalyst in the synthesis of:
  • 3,4-dihydropyrimidin-2(1H)-ones and thiones by reacting with aldehydes, β-ketoesters and urea/thiourea via one pot-three component Biginelli reaction.
  • Methyl methacryalate (MMA) by oxidative dehydrogenation of methyl iso-butarate (MIB).

It can be also used in the synthesis of carbon coated lithium iron phosphate (LiFePO4) as the cathode material for lithium ion batteries. It is also used as a key ingredient in preparing phosphate glass fibers.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Magnesium coated phosphate glass fibers for unidirectional reinforcement of polycaprolactone composites.
Liu X, et al.
Journal of Biomedical Materials Research. Part B, Applied Biomaterials, 103(7), 1424-1432 (2015)
Iron (III) phosphate dihydrate-catalyzed one-pot synthesis of dihydropyrimidinones and thiones: An improved procedure for the Biginelli reaction
Heravi MM, et al.
Heterocyclic Communications, 12(5), 369-372 (2006)
Optimized LiFePO4--polyacene cathode material for lithium-ion batteries.
Xie H_M, et al.
Advanced Materials, 18(19), 2609-2613 (2006)
Structural and electrochemical study of a new crystalline hydrated iron (III) phosphate FePO4
Marx N, et al.
Chemistry of Materials, 22(5), 1854-1861 (2010)
Yajing Yin et al.
Chemical communications (Cambridge, England), 48(15), 2137-2139 (2012-01-17)
Amorphous FePO(4) hollow nanospheres were directly grown on graphene for use as a cathode material in lithium ion batteries. This hybrid exhibits high rate capability and good cycle stability because of efficient Li(+) ion diffusion through the thin wall of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service