Skip to Content
Merck
All Photos(3)

Key Documents

338893

Sigma-Aldrich

Lithium phosphate

greener alternative

Synonym(s):

Lithiophosphate (Li3 PO4 ), Lithium orthophosphate (Li3 PO4 ), Trilithium monophosphate, Trilithium orthophosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Li3PO4
CAS Number:
Molecular Weight:
115.79
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
powder

grade

for analytical purposes

form

powder

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

concentration

25-27% NaOH (titration)

application(s)

battery manufacturing

greener alternative category

SMILES string

[Li+].[Li+].[Li+].[O-]P([O-])([O-])=O

InChI

1S/3Li.H3O4P/c;;;1-5(2,3)4/h;;;(H3,1,2,3,4)/q3*+1;/p-3

InChI key

TWQULNDIKKJZPH-UHFFFAOYSA-K

General description

Lithium phosphate (Li₃PO₄) is a white-colored powder with stringent quality-checking parameters to support R&D activities. Its primary use lies in battery technology, especially in the production of lithium iron phosphate (LiFePO₄) cathode materials for lithium-ion batteries. Beyond this, it is also employed in research activities in ceramics and glass manufacturing, catalysis, pharmaceuticals, optoelectronics, corrosion inhibition, laboratory reagents etc.

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Click here for more information.

Application

Lithium phosphate (Li₃PO₄) can be used as a precursor in the synthesis of lithium iron phosphate (LiFePO₄) cathode materials for lithium-ion batteries. It can be used as an electrolyte additive to enhance the performance and stability of lithium-ion batteries. Itcan be employed in the production of specialty glasses, where it improves the thermal and chemical properties of the glass.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Improving rate performance of LiFePO4 cathode materials by hybrid coating of nano-Li3PO4 and carbon
Zhao S, et al.
Journal of alloys and compounds, 566(11), 206-211 (2013)
Chemical vapor deposition of lithium phosphate thin-films for 3D all-solid-state Li-ion batteries
Xie J, et al.
Journal of the Electrochemical Society, 162(3), A249-A254 (2015)
A Reyes Jiménez et al.
Nanoscale, 10(4), 2128-2137 (2018-01-13)
In this work, we present a comprehensive study on the influence of lithium phosphorus oxynitride (LIPON) as a possible "artificial SEI layer" on the electrochemical performance of pure silicon (Si) thin film electrodes for a possible application in microbatteries or
Angel L Ortiz et al.
Dental materials : official publication of the Academy of Dental Materials, 36(5), 645-659 (2020-04-13)
To elucidate the compositional and microstructural developments of a novel lithium silicate glass-ceramic during its crystallization cycle. Blocks of a lithium silicate glass-ceramic (Obsidian®, Glidewell Laboratories) were cut into 1mm thick plates and polished to 1μm finish. Some of them
Synthesis and characterization of olivine LiNiPO4 for aqueous rechargeable battery
Minakshi M, et al.
Electrochimica Acta, 56(11), 4356-4360 (2011)

Articles

Innovation in dental restorative materials is driven by the need for biocompatible and natural-appearing restoration alternatives. Conventional dental materials like amalgam and composite resins have inherent disadvantages.

Lithium-ion batteries' characteristics make them popular for electricity storage due to portability, rechargeability, and low cost.

Lithium-ion batteries' characteristics make them popular for electricity storage due to portability, rechargeability, and low cost.

Lithium-ion batteries' characteristics make them popular for electricity storage due to portability, rechargeability, and low cost.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service