915432
Lithium manganese nickel oxide
spinel, powder, battery grade
Synonym(s):
LMNO, NANOMYTE® SP-10
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
Li2Mn3NiO8
CAS Number:
Molecular Weight:
365.38
MDL number:
UNSPSC Code:
12352303
NACRES:
NA.23
Recommended Products
grade
battery grade
Quality Level
description
Minimum Capacity: 115 mAh/g
Nominal Capacity (0.1C): ≥130 mAh/g
form
powder
mol wt
Mw 365.38 g/mol
composition
Li2Mn3NiO8
avg. part. size
5-7 μm (APS)
grain size
50-70 nm
density
4.27 g/cm3 (lit.)
application(s)
battery manufacturing
Related Categories
General description
Lithium manganese nickel oxide is a cathode material that can be charged at high voltage. Due to the high potential, the material has a higher energy density compared to lithium cobalt oxide and lithium iron phosphate.
Legal Information
NANOMYTE is a registered trademark of NEI Corporation
related product
Product No.
Description
Pricing
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Carc. 2 - Skin Sens. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Li-ion battery materials: present and future
N Nitta, et al.
Materials Today, 18(5), 252-264 (2015)
The lithium-ion battery: State of the art and future perspectives
G Zubi, et al.
Renewable and Sustainable Energy Reviews, 89, 292-308 (2018)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service