Skip to Content
Merck
All Photos(1)

Key Documents

745146

Sigma-Aldrich

9-Methyl-9H-fluorene-9-carbonyl-13C chloride

≥97.0% (GC)

Synonym(s):

13COgen

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13CC14H11ClO
CAS Number:
Molecular Weight:
243.69
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

isotopic purity

≥97.0%

Quality Level

Assay

≥97.0% (GC)

form

solid

reaction suitability

reaction type: C-C Bond Formation

SMILES string

CC1(c2ccccc2-c3ccccc13)[13C](Cl)=O

InChI

1S/C15H11ClO/c1-15(14(16)17)12-8-4-2-6-10(12)11-7-3-5-9-13(11)15/h2-9H,1H3/i14+1

InChI key

ZQYOOHGEBHBNTP-UJKGMGNHSA-N

Application

COGen is a simple alternative for carbon monoxide generation in the application of palladium-catalyzed carbonylation reactions. This reagent has proved to be applicable in a wide range of applications and is compatible with several building blocks in the formation of ketones, amides, esters, etc.

For more information please visit: Technology Spotlight, Professor Skrystrup PPP

Use with the COware Platform

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Linkage

Frequently Asked Questions are available for this Product.

also commonly purchased with this product

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[14C]-Carbon Monoxide
Journal of Labelled Compounds & Radiopharmaceuticals, 55, 411-418 (2012)
Philippe Hermange et al.
Organic letters, 13(9), 2444-2447 (2011-04-08)
A carbonylative Heck reaction of aryl iodides and styrene derivatives employing a two-chamber system using a stable, crystalline, and nontransition metal based carbon monoxide source is reported. By applying near-stoichiometric amounts of the carbon monoxide precursor, an effective exploitation of
Mia N Burhardt et al.
The Journal of organic chemistry, 77(12), 5357-5363 (2012-05-23)
We have synthesized two isotopically labeled variants of the β-amyloid binding compound FSB possessing (13)C-labels on the two terminal aryl carboxylic acid moieties. One of these was also fully deuterated on the olefinic spacers. The (13)C-isotope labeling was achieved applying
Korsager, S.;
Angewandte Chemie (International Edition in English) (2013)
Palladium-catalyzed carbonylative α-arylation for accessing 1,3-diketones.
Thomas M Gøgsig et al.
Angewandte Chemie (International ed. in English), 51(3), 798-801 (2011-12-06)

Articles

Studies in the field of carbonylation chemistry led to the discovery of a novel carbon monoxide (CO) delivery system.

Studies in the field of carbonylation chemistry led to the discovery of a novel carbon monoxide (CO) delivery system.

Studies in the field of carbonylation chemistry led to the discovery of a novel carbon monoxide (CO) delivery system.

Studies in the field of carbonylation chemistry led to the discovery of a novel carbon monoxide (CO) delivery system.

See All

Related Content

The Skrydstrup group has developed reagents and glassware for carrying out transition metal catalyzed carbonylations in a simple and safe manner.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service