205958
Platinum on carbon
extent of labeling: 10 wt. % loading, matrix activated carbon support
Synonym(s):
Pt/C
Sign Into View Organizational & Contract Pricing
All Photos(4)
About This Item
Empirical Formula (Hill Notation):
Pt
Molecular Weight:
195.08
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
form
powder
Quality Level
reaction suitability
core: platinum
reagent type: catalyst
greener alternative product characteristics
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
extent of labeling
10 wt. % loading
matrix
activated carbon support
greener alternative category
, Enabling
SMILES string
[Pt]
InChI
1S/Pt
InChI key
BASFCYQUMIYNBI-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.
Pt/C is used as a catalyst in the oxidation of glucose and gluconic acid. Catalyst deactivation occurs via oxygen chemisorption and platinum surface oxidation.
Pt/C is used as a catalyst in the oxidation of glucose and gluconic acid. Catalyst deactivation occurs via oxygen chemisorption and platinum surface oxidation.
Application
Catalyst used for the hydrogenation of alkenes, alkynes, nitro groups, ketones, aromatics; used for hydrogenolysis of cyclopropanes; catalyzes oxidation of alcohols.
Platinum on carbon (Pt/C) can be used as a catalyst for a variety of applications, such as:
- hydrosilylation reaction for the production of molecules with silicon-carbon bonds
- direct alcohol fuel cells (DAFCs)
- an electrocatalyst in hydrogen evolution reaction (HER)
- formation of nanocatalyst for polymer electrolyte fuel cells
Storage Class Code
11 - Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
The oxidation of gluconic acid with platinum on carbon as catalyst
Dirkx JM, et al.
J. Catal., 67, 14-20 (1981)
Oi, R.; Takenaka, S.
Chemistry Letters (Jpn), 7, 1115-1115 (1988)
Palmer, C. J.; Casida, J. E.
Tetrahedron Letters, 31, 2857-2857 (1990)
Platinum supported on mesoporous carbon as cathode catalyst for direct methanol fuel cells
Bruno MM, et al.
Journal of Power Sources, 278, 458-463 (2015)
Hydrosilylation of alkynes catalyzed by platinum on carbon.
Chauhan M, et al.
Journal of Organometallic Chemistry, 645(1), 1-13 (2002)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service