Skip to Content
Merck
All Photos(3)

Documents

738565

Sigma-Aldrich

Platinum cobalt on carbon

greener alternative

extent of labeling: 30 wt. % Pt3Co loading

Synonym(s):

27% Pt, 3% Co on High Durable Carbon, 30% Pt3Co/HDC

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CoPt3
Molecular Weight:
644.17
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.23

form

powder

Quality Level

reaction suitability

reagent type: catalyst
core: platinum

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

extent of labeling

30 wt. % Pt3Co loading

particle size

<5 nm (Pt)

greener alternative category

InChI

1S/Co.3Pt

InChI key

GUBSQCSIIDQXLB-UHFFFAOYSA-N

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 2 - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Siracusano, S.; Stassi, A.; Baglio, V.; Arico, A. S.; Capitanio, F.; Tavares, A. C.
Electrochimica Acta, 54, 4844-4844 (2009)
Antolini, E.
Materials Chemistry and Physics, 78, 563-563 (2003)

Articles

Proton exchange membrane (PEM) fuel cells operate at relatively low temperatures and are composed of two electrodes and a conductive elecrolyte.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service