Skip to Content
Merck
All Photos(2)

Key Documents

STW5

COware gas reactor

reaction scale of 1.0-5.0 mmol (total volume 100 mL)

Synonym(s):

Gas Reactor

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

UNSPSC Code:
41103817
NACRES:
NB.51

packaging

pack of 1 ea

Looking for similar products? Visit Product Comparison Guide

General description

Two-chamber glass apparatus are ideal for reaction scales of:

  • 0.1-1 mmol (total volume 20 mL)
  • 1.0-5.0 mmol (total volume 100 mL)
  • 5.0-20.0 mmol (total volume 400 mL)

Each unit includes two screw caps, two stabilizing PTFE discs (recyclable), ten single use silicone/PTFE septa (in combination with the stabilizing discs each septum allows for a maximum of 2 piercings using 21G/Ø0.80 mm syringes or smaller).

Order replacement PTFE/silicone septa STW3.

Learn more about CO releasing molecules from Prof. Skrydstrup


COware gas reactor is a part of the COware Platform

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Studies in the field of carbonylation chemistry led to the discovery of a novel carbon monoxide (CO) delivery system.

Studies in the field of carbonylation chemistry led to the discovery of a novel carbon monoxide (CO) delivery system.

Studies in the field of carbonylation chemistry led to the discovery of a novel carbon monoxide (CO) delivery system.

Studies in the field of carbonylation chemistry led to the discovery of a novel carbon monoxide (CO) delivery system.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service