Skip to Content
Merck
All Photos(1)

Documents

740306

Sigma-Aldrich

Vitamin D2 solution

100 μg/mL in ethanol, 98% (CP)

Synonym(s):

Calciferol solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H44O
Molecular Weight:
396.65
EC Number:
MDL number:
UNSPSC Code:
12352200
eCl@ss:
34058017
PubChem Substance ID:
NACRES:
NA.24

Assay

98% (CP)

form

solution

concentration

100 μg/mL in ethanol

technique(s)

mass spectrometry (MS): suitable

color

colorless

application(s)

detection

storage temp.

−20°C
−20°C

SMILES string

CC(C)[C@@H](C)\C=C\[C@@H](C)[C@H]1CC[C@H]2\C(CCC[C@]12C)=C\C=C3\C[C@@H](O)CCC3=C

InChI

1S/C28H44O/c1-19(2)20(3)9-10-22(5)26-15-16-27-23(8-7-17-28(26,27)6)12-13-24-18-25(29)14-11-21(24)4/h9-10,12-13,19-20,22,25-27,29H,4,7-8,11,14-18H2,1-3,5-6H3/b10-9+,23-12+,24-13-/t20-,22+,25-,26+,27-,28+/m0/s1

InChI key

MECHNRXZTMCUDQ-RKHKHRCZSA-N

Gene Information

human ... VDR(7421)

Related Categories

General description

Vitamin D2 is also known as ergocalciferol. It is produced by plants and mushrooms using sunlight.

Application

Vitamin D2 solution (Calciferol solution) can be used as an internal standard for the quantification of vitamin D in biological samples by a liquid chromatography-tandem mass spectrometry (LC-MS/MS).

Packaging

1 mL in ampule

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

55.4 °F

Flash Point(C)

13 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous determination of cholecalciferol (vitamin D3) and ergocalciferol (vitamin D2) in foods by selected reaction monitoring
Dimartino G
Journal of AOAC (Association of Official Analytical Chemists) International, 92(2), 511-517 (2009)
Sejal Shah et al.
The Journal of adolescent health : official publication of the Society for Adolescent Medicine, 57(1), 19-23 (2015-04-16)
Vitamin D deficiency and cardiometabolic risk factors are common in obese adolescents. Observational studies demonstrate an inverse relationship among serum 25-hydroxyvitamin D (25OHD) and obesity, insulin resistance, and inflammatory cytokines. This pilot study explores if vitamin D supplementation could reduce
Sterol and vitamin D2 concentrations in cultivated and wild grown mushrooms: Effects of UV irradiation
Teichmann A, et al.
LWT--Food Science and Technology, 40(5), 815-822 (2007)
Tom D Thacher et al.
Archives of disease in childhood, 99(9), 807-811 (2014-04-22)
To determine whether children with calcium-deficiency rickets have a better response to treatment with vitamin D and calcium than with calcium alone. Randomised controlled trial. Jos University Teaching Hospital, Jos, Nigeria. Nigerian children with active rickets treated with calcium carbonate
James E Heubi et al.
Hepatology (Baltimore, Md.), 61(1), 268-274 (2014-08-29)
Bile acid amidation defects were predicted to present with fat/fat soluble vitamin malabsorption with minimal cholestasis. We identified and treated five patients (one male, four females) from four families with defective bile acid amidation due to a genetically confirmed deficiency

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service