Skip to Content
Merck

Skip To

V-024

Vitamin D2 solution

1 mg/mL in ethanol, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

Vitamin D2 solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C28H44O
CAS Number:
Molecular Weight:
396.65
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

form

liquid

feature

(Snap-N-Shoot®), (Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1 mg/mL in ethanol

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

mp

-144.0 °C

application(s)

clinical testing

format

single component solution

storage temp.

−70°C

SMILES string

O[C@H]1CCC(=C)\C(=C/C=C2/[C@H]3[C@@]([C@H](CC3)[C@H](C)\C=C\[C@@H](C(C)C)C)(CCC/2)C)\C1

InChI

1S/C28H44O/c1-19(2)20(3)9-10-22(5)26-15-16-27-23(8-7-17-28(26,27)6)12-13-24-18-25(29)14-11-21(24)4/h9-10,12-13,19-20,22,25-27,29H,4,7-8,11,14-18H2,1-3,5-6H3/b10-9+,23-12+,24-13-/t20-,22+,25-,26+,27-,28+/m0/s1

InChI key

MECHNRXZTMCUDQ-RKHKHRCZSA-N

Gene Information

human ... VDR(7421)

General description

Vitamin D2 is a form of vitamin D also known as ergocalciferol. Marketed under trade names including Deltalin, Drisdol®, and Calcidol®, vitamin D2 is often used as a vitamin D supplement for individuals with vitamin D deficiency.

Application

This certified solution standard can be used Snap-N-Spike® as a starting material in the preparation of calibrators or controls for clinical applications or Snap-N-Shoot® in quality control testing of vitamin supplements, fortified foods or pharmaceutical preparations.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Calcidol is a registered trademark of Patrin Pharma, Inc.
Drisdol is a registered trademark of Sanofi-Aventis
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHR1238E0900000E5750
technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

technique(s)

HPLC: suitable

application(s)

clinical testing

application(s)

clinical testing

application(s)

pharmaceutical (small molecule)

application(s)

-

form

liquid

form

-

form

solid

form

powder

format

single component solution

format

neat

format

neat

format

-

Quality Level

300

Quality Level

300

Quality Level

-

Quality Level

200

grade

certified reference material

grade

certified reference material, pharmaceutical secondary standard

grade

pharmaceutical primary standard

grade

-


signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT RE 2 Oral

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

57.2 °F

flash_point_c

14.0 °C



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Sejal Shah et al.
The Journal of adolescent health : official publication of the Society for Adolescent Medicine, 57(1), 19-23 (2015-04-16)
Vitamin D deficiency and cardiometabolic risk factors are common in obese adolescents. Observational studies demonstrate an inverse relationship among serum 25-hydroxyvitamin D (25OHD) and obesity, insulin resistance, and inflammatory cytokines. This pilot study explores if vitamin D supplementation could reduce
Tom D Thacher et al.
Archives of disease in childhood, 99(9), 807-811 (2014-04-22)
To determine whether children with calcium-deficiency rickets have a better response to treatment with vitamin D and calcium than with calcium alone. Randomised controlled trial. Jos University Teaching Hospital, Jos, Nigeria. Nigerian children with active rickets treated with calcium carbonate
James E Heubi et al.
Hepatology (Baltimore, Md.), 61(1), 268-274 (2014-08-29)
Bile acid amidation defects were predicted to present with fat/fat soluble vitamin malabsorption with minimal cholestasis. We identified and treated five patients (one male, four females) from four families with defective bile acid amidation due to a genetically confirmed deficiency



Global Trade Item Number

SKUGTIN
V-024-1ML04061837428630

Questions

Reviews

No rating value

Active Filters