Skip to Content
Merck
All Photos(1)

Key Documents

527750

Sigma-Aldrich

4-(Hydroxymethyl)benzonitrile

95%

Synonym(s):

4-Cyanobenzyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2C6H4CN
CAS Number:
Molecular Weight:
133.15
Beilstein:
2206193
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

95%

mp

39-43 °C (lit.)

functional group

hydroxyl
nitrile

SMILES string

OCc1ccc(cc1)C#N

InChI

1S/C8H7NO/c9-5-7-1-3-8(6-10)4-2-7/h1-4,10H,6H2

InChI key

XAASLEJRGFPHEV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-(Hydroxymethyl)benzonitrile can be synthesized via hydrosilylation reaction in the presence of Fe complex Bu4N[Fe(CO)3(NO)] [catalyst].[1] It can also be prepared from 4-(aminomethyl) benzyl alcohol.[2]

Application

4-(Hydroxymethyl)benzonitrile [p-(hydroxymethyl)benzonitrile] may be used to synthesize p-((vinyloxy)methyl)benzonitrile (VOMBN).[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bergstra?er U, et al.
Science of Synthesis: Houben-Weyl Methods of Molecular Transformations, 219-219 (2014)
Vinyl monomers bearing chromophore moieties and their polymers. VIII. synthesis and fluorescence behavior of a vinyloxy monomer having an electron-accepting chromophore moiety, p-((vinyloxy) methyl) benzonitrile, and its polymers.
Du FS, et al.
Journal of Polymer Science Part A: Polymer Chemistry, 37(2), 179-187 (1999)
Bu4N [Fe (CO)3(NO)]-Catalyzed Hydrosilylation of Aldehydes and Ketones.
Dieskau AP, et al.
European Journal of Organic Chemistry, 27, 5291-5296 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service