Skip to Content
Merck
All Photos(5)

Key Documents

139009

Sigma-Aldrich

1,8-Diazabicyclo[5.4.0]undec-7-ene

greener alternative

98%

Synonym(s):

2,3,4,6,7,8,9,10-Octahydropyrimidol[1,2-a]azepine, DBU

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H16N2
Molecular Weight:
152.24
Beilstein:
508906
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

5.3 mmHg ( 37.7 °C)

Quality Level

Assay

98%

form

liquid

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

refractive index

n20/D 1.522-1.524 (lit.)

bp

80-83 °C/0.6 mmHg (lit.)

mp

-70 °C

density

1.018 g/mL at 25 °C (lit.)

greener alternative category

SMILES string

C1CCN2CCCN=C2CC1

InChI

1S/C9H16N2/c1-2-5-9-10-6-4-8-11(9)7-3-1/h1-8H2

InChI key

GQHTUMJGOHRCHB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,8-Diazabicyclo[5.4.0]undec-7-ene is a bicyclic amidine base. It is non-nucleophilic, sterically hindered, tertiary amine base in organic chemistry. It is reported to be superior to amine catalyst in Baylis-Hillman reaction. It promotes the methylation reaction of phenols, indoles and benzimidazoles with dimethyl carbonate under mild conditions.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for catalytic efficiency. Click here for more information.

Application

1,8-Diazabicyclo[5.4.0]undec-7-ene (DBU) may be used:
  • as catalyst for carboxylic acid esterification with dimethyl carbonate
  • in the synthesis of duocarmycin and CC-1065 analogs
  • as catalyst in aza-Michael addition and Knovenegal condensation reaction
  • as base for dehalogenation of halogenated Diels-Alder adducts and the resulting activated 2,4-dienones were subjected to regio- and stereo-directed Michael additions, using Yamamoto′s reagent (CH3Cu · BF3)
  • in a new synthesis of the ABCD ring system of Camptothecin
1,8-Diazabicyclo[5.4.0]undec-7-ene may be used as an catalyst for the dissolution and activation of cellulose by a reversible reaction of its hydroxyl groups with carbon dioxide. This dissolved cellulose system can be derivatized to form cellulose mixed esters.
Used in a new synthesis of the ABCD ring system of Camptothecin.

Features and Benefits

Strong hindered amine base.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

240.8 °F

Flash Point(C)

116 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

DBU (1, 8-diazabicyclo [5.4. 0] undec-7-ene)-A Nucleophillic Base.
Ghosh N
Synlett, 2004(03), 574-575 (2004)
Zhi-Hao Cui et al.
Journal of chemical theory and computation, 16(1), 119-129 (2019-12-10)
We describe an efficient quantum embedding framework for realistic ab initio density matrix embedding theory (DMET) calculations in solids. We discuss in detail the choice of orbitals and mapping to a lattice, treatment of the virtual space and bath truncation
Sustainable succinylation of cellulose in a CO 2-based switchable solvent and subsequent Passerini 3-CR and Ugi 4-CR modification.
Soyler Z, et al.
Green Chemistry, 20(1), 214-224 (2018)
Synthesis of cellulose acetate propionate and cellulose acetate butyrate in a CO2/DBU/DMSO system.
Xu Q, et al.
Cellulose, 25(1), 205-216 (2018)
Weixiang Dai et al.
Organic letters, 8(20), 4665-4667 (2006-09-22)
A new synthetic pathway to the parent and substituted ABCD ring cores of the camptothecin family of alkaloids was developed. The N-alkylation of 1,6-dihydro-6-oxo-2-pyridinecarbonitrile (2) with 3-bromo-1-phenylpropyne provided 3a using Curran's protocol. Treatment of 3a with a catalytic amount of

Articles

The prevalence of organofluorine compounds in industry and drug design necessitates the ability to introduce C–F bonds to molecules.

ASIF provides bench-stable alternative to sulfuryl fluoride gas for installing SO2F functional group in organic synthesis.

ASIF provides bench-stable alternative to sulfuryl fluoride gas for installing SO2F functional group in organic synthesis.

ASIF provides bench-stable alternative to sulfuryl fluoride gas for installing SO2F functional group in organic synthesis.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service