Skip to Content
Merck
All Photos(1)

Key Documents

132330

Sigma-Aldrich

p-Tolunitrile

98%

Synonym(s):

4-Methylbenzonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4CN
CAS Number:
Molecular Weight:
117.15
Beilstein:
507386
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

solid

bp

103-106 °C/20 mmHg (lit.)
217 °C (lit.)

mp

26-28 °C (lit.)

solubility

alcohol: very soluble
diethyl ether: very soluble
water: insoluble

density

0.981 g/mL at 25 °C (lit.)

functional group

nitrile

SMILES string

Cc1ccc(cc1)C#N

InChI

1S/C8H7N/c1-7-2-4-8(6-9)5-3-7/h2-5H,1H3

InChI key

VCZNNAKNUVJVGX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Kei Ohkubo et al.
Journal of the American Chemical Society, 125(42), 12850-12859 (2003-10-16)
A ring-substituted toluene with an electron-withdrawing substituent, p-tolunitrile, is oxygenated by molecular oxygen to yield the corresponding aldehyde with tetrafluoro-p-dicyanobenzene as a photocatalyst under photoirradiation with an Hg lamp (lambda > 300 nm). The oxygenation of a ring-substituted toluene with
Vibronic spectroscopy of jet-cooled p-cyanobenzyl radical: D1? D0 spectra and vibrational mode assignments.
Lee SK and Ahn BU.
Chemical Physics Letters, 320(5), 601-606 (2000)
Niina Tani et al.
Bioorganic & medicinal chemistry, 22(23), 6655-6664 (2014-12-03)
Inhibition of CYP2A6-mediated nicotine metabolism can reduce cigarette smoking. We sought potent and selective CYP2A6 inhibitors to be used as leads for drugs useful in smoking reduction therapy, by evaluating CYP2A6 inhibitory effect of novel formyl, alkyl amine or carbonitrile

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service