Skip to Content
Merck
All Photos(1)

Key Documents

146250

Sigma-Aldrich

3-Formylbenzonitrile

98%

Synonym(s):

3-Cyanobenzaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NCC6H4CHO
CAS Number:
Molecular Weight:
131.13
Beilstein:
2205751
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

powder

bp

210 °C (lit.)

mp

75-78 °C (lit.)

SMILES string

[H]C(=O)c1cccc(c1)C#N

InChI

1S/C8H5NO/c9-5-7-2-1-3-8(4-7)6-10/h1-4,6H

InChI key

HGZJJKZPPMFIBU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3-Formylbenzonitrile was used in the synthesis of 3-(6,6-dimethyl-5,6-dihydro-4H-benzo[7,8]chromeno[6,5-d]oxazol-2-yl)benzonitrile.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kelly C G Moura et al.
Bioorganic & medicinal chemistry, 20(21), 6482-6488 (2012-09-25)
Twenty-three naphthoimidazoles and six naphthoxazoles were synthesised and evaluated against susceptible and rifampicin- and isoniazid-resistant strains of Mycobacterium tuberculosis. Among all the compounds evaluated, fourteen presented MIC values in the range of 0.78 to 6.25 μg/mL against susceptible and resistant
Hong Zhao et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 10(10), 2386-2390 (2004-05-18)
The molten reaction of 2-naphthol, 4-(aminomethyl)pyridine, and 4-pyridinecarboxaldehyde at about 180 degrees C yields trans-2,3-dihydro-2,3-di(4'-pyridyl)benzo[e]indole (1) which possesses two chiral centers, rather than an expected Betti-type reaction product with only one chiral carbon center. The same reactions, using 3-pyridinecarboxaldehyde, 4-cyanobenzaldehyde

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service