Skip to Content
Merck
All Photos(1)

Key Documents

367729

Sigma-Aldrich

Tridodecylmethylammonium chloride

98%

Synonym(s):

Methyltridodecylammonium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)11]3N(Cl)CH3
CAS Number:
Molecular Weight:
572.47
Beilstein:
6031445
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

110-112 °C (lit.)

SMILES string

[Cl-].CCCCCCCCCCCC[N+](C)(CCCCCCCCCCCC)CCCCCCCCCCCC

InChI

1S/C37H78N.ClH/c1-5-8-11-14-17-20-23-26-29-32-35-38(4,36-33-30-27-24-21-18-15-12-9-6-2)37-34-31-28-25-22-19-16-13-10-7-3;/h5-37H2,1-4H3;1H/q+1;/p-1

InChI key

SBHRWOBHKASWGU-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Tridodecylmethylammonium chloride can be used as a:
  • Catalyst in the transesterification of methyl esters with glycidol to synthesize glycidyl esters.
  • Bulky lipophilic cationic additive for polyaniline membranes.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quaternary alkyl ammonium salt-catalyzed transformation of glycidol to glycidyl esters by transesterification of methyl esters
Tanaka S, et al.
ACS Catalysis, 8, 1097-1103 (2018)
Multisensing ion-selective field-effect transistors prepared by ionophore doping technique.
K Bezegh et al.
Analytical chemistry, 59(24), 2846-2848 (1987-12-15)
Raman and UV-vis spectroscopic study of polyaniline membranes containing a bulky cationic additive
Lindfors T, et al.
Journal of Electroanalytical Chemistry, 518, 131-138 (2002)
Andrew Yu et al.
Analytica chimica acta, 686(1-2), 76-80 (2011-01-18)
The development of a polymer membrane-based electrode to measure the anionic drug suramin in buffered saline and biological samples is described. A large non-equilibrium, steady state EMF response is observed toward suramin, and judicious choice of the polymer membrane components
Natalia Abramova et al.
Talanta, 81(4-5), 1750-1754 (2010-05-06)
The influence of CO(2) and acetic acid on the response of ISFET sensors with PVC and photocured polyurethane polymer membranes with valinomycin as an ionophore was assessed. Experimental results show that the presence of these compounds has no effect on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service