Skip to Content
Merck
All Photos(1)

Key Documents

358711

Sigma-Aldrich

Tetraethylammonium cyanide

94%

Synonym(s):

N,N,N-triethyl-ethanaminium cyanide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)4N(CN)
CAS Number:
Molecular Weight:
156.27
Beilstein:
3570222
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Assay

94%

form

powder

mp

250 °C (dec.) (lit.)

SMILES string

[C-]#N.CC[N+](CC)(CC)CC

InChI

1S/C8H20N.CN/c1-5-9(6-2,7-3)8-4;1-2/h5-8H2,1-4H3;/q+1;-1

InChI key

PCZOZSATUTWXIC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Tetraethylammonium cyanide can be used:
  • As a cyanide source for 1,4-addition of cyano group to enones (chalcones) catalyzed by scandium(III) triflate to yield β-cyanoketones.
  • Along with diethylazodicarboxylate (DEAD) in the regioselective C- or N- ethoxy-carbonylation of heterocyclic systems like indoles and indazoles.
  • In the synthesis of C-C-bridged bis-isoflavones from 2-bromomethyl-7,4′-dimethoxyisoflavone.

Tetraethylammonium cyanide is a versatile reagent commonly used in cyanation reactions as cyanide source. It is also used in the preparation of various cyanometalate complexes.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Supplementary Hazards

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Synthesis of C- C-bridged bis-isoflavones
Al-Maharik NI, et al.
The Journal of Organic Chemistry, 65(8), 2305-2308 (2000)
An S= 2 cyanide-bridged trinuclear FeIII2NiII single-molecule magnet.
Li D, et al.
Inorganic Chemistry, 45(14), 5251-5253 (2006)
Tetraethylammonium Cyanide.
e-EROS Encyclopedia of Reagents for Organic Synthesis (2009)
Nucleophilic aromatic substitution of hydrogen: a novel electrochemical approach to the cyanation of nitroarenes.
Gallardo I, et al.
Chemistry?A European Journal , 7(8), 1759-1765 (2001)
Regioselective ethoxy-carbonylation of indoles and indazoles using DEAD and tetraethylammonium cyanide
Ramesh S, et al.
Royal Society of Chemistry Advances, 3(23), 8666-8669 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service