Skip to Content
Merck
All Photos(1)

Key Documents

212849

Sigma-Aldrich

Trimethylsilyl cyanide

98%

Synonym(s):

Cyanotrimethylsilane, TMSCN

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3SiCN
CAS Number:
Molecular Weight:
99.21
Beilstein:
1737612
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.392 (lit.)

bp

114-117 °C (lit.)

mp

8-11 °C (lit.)

density

0.793 g/mL at 20 °C (lit.)

SMILES string

C[Si](C)(C)C#N

InChI

1S/C4H9NSi/c1-6(2,3)4-5/h1-3H3

InChI key

LEIMLDGFXIOXMT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Trimethylsilyl cyanide participates in carbonyl aminomethylation via α-silyloxy nitriles.

Application

Trimethylsilyl cyanide was used:
  • for derivatization of complex metabolite mixtures by GC-MS
  • as cyanide source in enantioselective organocatalytic Strecker-type reaction of aliphatic N,N-dialkylhydrazones
  • as reagent for the cyanosilylation of aldehydes in near quantitative yield via catalysis by silylene-bridged rare earth oxide complexes
  • in asymmetric cyanosilylation with chiral dinuclear Ti(IV) complexes

Packaging

Packaging material contains granular sodium carbonate as a desiccant.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Flam. Liq. 2

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

33.8 °F - closed cup

Flash Point(C)

1 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthetic Communications, 36, 2483-2483 (2006)
Bekzod Khakimov et al.
Analytical and bioanalytical chemistry, 405(28), 9193-9205 (2013-10-05)
Reproducible and quantitative gas chromatography-mass spectrometry (GC-MS)-based metabolomics analysis of complex biological mixtures requires robust and broad-spectrum derivatization. We have evaluated derivatization of complex metabolite mixtures using trimethylsilyl cyanide (TMSCN) and the most commonly used silylation reagent N-methyl-N-(trimethylsilyl)trifluoroacetamide (MSTFA). For
Tetrahedron Asymmetry, 17, 2328-2328 (2006)
Aurora Martínez-Muñoz et al.
Organic & biomolecular chemistry, 11(47), 8247-8255 (2013-10-30)
The enantioselective organocatalytic Strecker-type reaction of aliphatic N,N-dialkylhydrazones is presented. Using trimethylsilyl cyanide (TMSCN) as the cyanide source, the reaction can be efficiently catalyzed by a tert-leucine-derived bifunctional thiourea to afford the corresponding hydrazino nitriles in good to excellent yields
Synthetic applications of trimethylsilyl cyanide. Efficient synthesis of. beta.-aminomethyl alcohols.
Evans DA, et al.
The Journal of Organic Chemistry, 39(7), 914-917 (1974)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service