Skip to Content
Merck
All Photos(1)

Key Documents

164283

Sigma-Aldrich

Methyl trifluoromethanesulfonate

≥98%

Synonym(s):

Methyl triflate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3SO2OCH3
CAS Number:
Molecular Weight:
164.10
Beilstein:
774772
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98%

form

liquid

refractive index

n20/D 1.326 (lit.)

bp

94-99 °C (lit.)

density

1.45 g/mL at 25 °C (lit.)

functional group

fluoro
triflate

storage temp.

2-8°C

SMILES string

COS(=O)(=O)C(F)(F)F

InChI

1S/C2H3F3O3S/c1-8-9(6,7)2(3,4)5/h1H3

InChI key

OIRDBPQYVWXNSJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methyl trifluoromethanesulfonate is a strong methylating reagent, commonly used for the pre-methylation of polysaccharides under mild basic conditions.

Application

Methyl trifluoromethanesulfonate can be used as a methylation reagent:
  • In the determination of polysulfides, zerovalent sulfur in sulfide-rich water wells, and polysulfide species in electrolyte of a lithium–sulfur battery using chromatography-based techniques.
  • In reactions with potassium enolates.
  • For the conversion of amines to methyl ammonium triflates.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

100.4 °F - closed cup

Flash Point(C)

38 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oliver Schuster et al.
Inorganic chemistry, 45(20), 7997-7999 (2006-09-27)
Two cationic carbene complexes with no heteroatom in the ring containing the carbene carbon, trans-bromo(2-methyl-2,6-dihydroisoquinolin-6-ylidene)bis(triphenylphosphine)palladium(II) triflate (3) and trans-chloro(1,2-dimethyl-1,7-dihydroquinolin-7-ylidene)bis(triphenylphosphine)palladium(II) triflate (4), were synthesized by oxidative substitution of Pd(PPh3)4 with N-methylated 6-bromoisoquinolinium and 7-chloro-2-methylquinolinium cations, respectively. Compound 3 was also prepared
Ring Expansions of 2-Alkenylazetidinium Salts-a New Route to Pyrrolidines and Azepanes
Couty F, et al.
European Journal of Organic Chemistry, 2006(18), 4214-4223 (2006)
Quantitative and Qualitative Determination of Polysulfide Species in the Electrolyte of a Lithium?Sulfur Battery using HPLC ESI/MS with One?Step Derivatization
Zheng D, et al.
Advanced Energy Materials, 5(16) (2015)
Purification of Laboratory Chemicals, 6 (2009)
Analytical Methods in Wood Chemistry, Pulping, and Papermaking (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service