Skip to Content
Merck
All Photos(2)

Key Documents

175501

Sigma-Aldrich

Boron trifluoride diethyl etherate

for synthesis

Synonym(s):

Boron trifluoride ethyl etherate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BF3 · O(C2H5)2
CAS Number:
Molecular Weight:
141.93
Beilstein:
3909607
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

grade

for synthesis
synthesis grade

Quality Level

vapor density

4.9 (vs air)

vapor pressure

4.2 mmHg ( 20 °C)

form

liquid

expl. lim.

36 %

reaction suitability

core: boron
reagent type: Lewis acid
reagent type: catalyst

refractive index

n20/D 1.344 (lit.)

bp

126-129 °C (lit.)

mp

−58 °C (lit.)

density

1.15 g/mL (lit.)

functional group

ether

storage temp.

2-8°C

SMILES string

CC[O+](CC)[B-](F)(F)F

InChI

1S/C4H10BF3O/c1-3-9(4-2)5(6,7)8/h3-4H2,1-2H3

InChI key

MZTVMRUDEFSYGQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Boron trifluoride diethyl etherate or boron trifluoride-ether complex is an organic compound widely used as a convenient source of boron trifluoride (BF3) in organic synthesis. It is also used as a Lewis acid for the activation of electrophiles.

Application

Catalyst used in the preparation of cyclopentyl- and cycloheptyl[b]indoles from aryl cyclopropyl ketones via [3+2] cycloaddition.
Lewis acid reagent with broad application

Packaging

The 25 mL Sure/Seal bottle is recommended as a single-use bottle. Repeated punctures will likely result in decreased performance of product.

Legal Information

Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B - STOT RE 1 Inhalation

Target Organs

Kidney

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

137.3 °F - closed cup

Flash Point(C)

58.5 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

European Journal of Organic Chemistry, 5378-5378 (2006)
Yong-Gang Xiang et al.
Chemical communications (Cambridge, England), (45)(45), 7045-7047 (2009-11-12)
The synergistic action of BF(3).OEt(2) and SmI(2) allowed a series of intermolecular cross-couplings of readily available N-acyl N,O-acetals with alpha,beta-unsaturated compounds to be performed in high yields, which was applied to the stereoselective synthesis of pyrrolizidine alkaloid (+)-xenovenine.
Devendra J Vyas et al.
The Journal of organic chemistry, 75(19), 6720-6723 (2010-09-10)
The BF(3)·OEt(2)-mediated allylation of isatin with an α-chiral allylic stannane is diastereo- and enantioselective. Conversely, allylation of any substituted isatin employing the identical protocol is not diastereoselective at all and only enantioselective for the major diastereomer having syn relative configuration.
Brajendra K Sharma et al.
Journal of agricultural and food chemistry, 56(9), 3049-3056 (2008-04-11)
Vegetable oils are promising candidates as substitutes for petroleum base oils in lubricant applications, such as total loss lubrication, military applications, and outdoor activities. Although vegetable oils have some advantages, they also have poor oxidation and low temperature stability. One
Bjart Frode Lutnaes et al.
Journal of the American Chemical Society, 126(29), 8981-8990 (2004-07-22)
A series of charge-delocalized carotenoid mono- and dications have been prepared by treatment of selected carotenoids with Brønsted and Lewis acids. The detailed structures of the carbocations were established by NMR studies in the temperature range from -10 to -20

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service