Skip to Content
Merck
All Photos(1)

Key Documents

357960

Sigma-Aldrich

1-Isoquinolinyl phenyl ketone

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H11NO
CAS Number:
Molecular Weight:
233.26
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

mp

76-79 °C (lit.)

functional group

ketone
phenyl

SMILES string

O=C(c1ccccc1)c2nccc3ccccc23

InChI

1S/C16H11NO/c18-16(13-7-2-1-3-8-13)15-14-9-5-4-6-12(14)10-11-17-15/h1-11H

InChI key

MDWCIKACMBMJFA-UHFFFAOYSA-N

General description

1-Isoquinolinyl phenyl ketone (iquinpk) reacts with ReCl3(MeCN)(PPh3)] complex to give [ReCl3(N-O)(PPh3)]. It also reacts with the [ReOBr3(PPh3)2] complex in acetone to yield novel complex, [ReOBr2(iquinpk-OH)(PPh3)].

Application

1-Isoquinolinyl phenyl ketone may be used in chemical synthesis.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, spectroscopic characterization, X-ray structure and DFT calculations of rhenium (III) complex with 1-isoquinolinyl phenyl ketone.
Machura B, et al.
Polyhedron, 27(6), 1739-1746 (2008)
Nucleophilic addition of water to 1-isoquinolinyl phenyl ketone. The synthesis, spectroscopic investigation, crystal and molecular structure and DFT calculations of [ReOBr2(iquinpk-OH)(PPh3)].
Machura,B, et al.
Inorganic Chemistry Communications, 12(8), 789-792 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service