Skip to Content
Merck
All Photos(2)

Key Documents

301884

Sigma-Aldrich

Phenyl 2-pyridyl ketoxime

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H10N2O
CAS Number:
Molecular Weight:
198.22
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

powder

mp

154-156 °C (lit.)

functional group

amine
oxime
phenyl

SMILES string

O\N=C(/c1ccccc1)c2ccccn2

InChI

1S/C12H10N2O/c15-14-12(10-6-2-1-3-7-10)11-8-4-5-9-13-11/h1-9,15H/b14-12+

InChI key

RSJDEVMJZLLAHS-WYMLVPIESA-N

General description

The complexation reaction of phenyl 2-pyridyl ketoxime with some transition and heavy metal ions Co2+, Ni2+, Zn2+, Pb2+, Fe2+, Fe3+, Cr3+ and La2+ has been studied potentiometrically. The reactions of phenyl(2-pyridyl)ketone oxime, with nickel(II) sulfate hexahydrate, in the absence of an external base, have been investigated.

Application

Phenyl 2-pyridyl ketoxime was used in preparation of inverse-9-metallacrown-3, trinuclear clusters.

Quality

May exhibit syn and anti forms

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Constantina Papatriantafyllopoulou et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 70(4), 718-728 (2007-10-10)
The reactions of phenyl(2-pyridyl)ketone oxime (py)C(ph)NOH, with nickel(II) sulfate hexahydrate under reflux, in the absence of an external base, have been investigated. The reaction of NiSO4 x 6H2O with two equivalents of (py)C(ph)NOH in H2O/MeOH leads to the dinuclear complex
Tereza Afrati et al.
Dalton transactions (Cambridge, England : 2003), 39(3), 765-775 (2010-01-13)
Interaction of phenyl 2-pyridyl ketoxime (PhPyCNO)/X(-)"blends" (X(-) = OH(-), Cl(-), ClO(4)(-), 2,4-D) (2,4-DH = 2,4-dichlorophenoxyacetic acid) with copper perchlorate has yielded trinuclear clusters that have been characterized as inverse-9-metallacrown-3, accommodating one or two anions. The structure of the complex [Cu(3)(PhPyCNO)(3)(mu(3)-OH)(2,4-D)(2)]
Ardeshir Shokrollahi et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 71(2), 655-662 (2008-03-11)
The complexation reaction between some oximes including methyl-2-pyridylketone oxime (MPKO), phenyl-2-pyridylketone oxime (PPKO) and diacetyl monooxime (DMO) with some transition and heavy metal ions Co2+, Ni2+, Zn2+, Pb2+, Fe2+, Fe3+, Cr3+ and La3+ has been studied potentiometrically in aqueous solution

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service