Skip to Content
Merck
All Photos(1)

Documents

238309

Sigma-Aldrich

Hexafluoroacetylacetone

98%

Synonym(s):

1,1,1,5,5,5-Hexafluoro-2,4-pentanedione, 1,1,1,5,5,5-Hexafluoroacetylacetone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3COCH2COCF3
CAS Number:
Molecular Weight:
208.06
Beilstein:
1790138
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.332 (lit.)

bp

70-71 °C (lit.)

density

1.47 g/mL at 25 °C (lit.)

SMILES string

FC(F)(F)C(=O)CC(=O)C(F)(F)F

InChI

1S/C5H2F6O2/c6-4(7,8)2(12)1-3(13)5(9,10)11/h1H2

InChI key

QAMFBRUWYYMMGJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Hexafluoroacetylacetone forms metal-chelate complexes with Cu(II), Ni(II), Co(II), Nd(III), Rh(III), Fe(III) and Cr(III). Hexafluoroacetylacetone on hydration yields 1,1,1,5,5,5-hexafluoropentane-2,2,4,4-tetraol.

Application


  • Antipsychotic medications and sleep problems in patients with schizophrenia: This research investigates the effects of antipsychotics, including aripiprazole, on sleep disturbances in schizophrenia patients, highlighting its pharmacodynamic actions and contributing to a deeper understanding of aripiprazole′s clinical applications in mental health disorders (Cederlöf et al., 2024).

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

89.6 °F - closed cup

Flash Point(C)

32 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Infrared Spectra of Metal Chelate Compounds of Hexafluoroacetylacetone.
Morris ML, et al.
Inorganic Chemistry, 2(2), 411-412 (1963)
A new fluorinated tetraalkoxide ligand derived from the hydration of hexafluoroacetylacetone.
Bouwman E, et al.
Inorganic Chemistry, 31(22), 4436-4438 (1992)
Zubair Ahmed et al.
Dalton transactions (Cambridge, England : 2003), 48(15), 4973-4986 (2019-03-28)
This paper presents three crystallographically and solution NMR (one- and two-dimensional) characterised ternary Ln(iii) complexes, and their photo-physical and opto-electronic properties in the visible and NIR regions. The single crystal X-ray diffraction analysis indicates that the complexes are nona-coordinate with
Shayan Shahbazi et al.
Journal of radioanalytical and nuclear chemistry, 311(1), 617-626 (2017-01-24)
This work reports the thermodynamic characterizations of organometallic species as a vehicle for the rapid separation of volatile nuclear fission products via gas chromatography due to differences in adsorption enthalpy. Because adsorption and sublimation thermodynamics are linearly correlated, there is
Yuichi Kitagawa et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 17(6), 845-849 (2016-01-21)
Magneto optical devices based on the Faraday effects of lanthanide ion have attracted much attention. Recently, large Faraday effects were found in nano-sized multinuclear lanthanide complexes. In this study, the Faraday rotation intensities were estimated for lanthanide nitrates [Ln(III) (NO3

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service