Skip to Content
Merck
All Photos(3)

Key Documents

323411

Sigma-Aldrich

Potassium tetrachloroplatinate(II)

99.99% trace metals basis

Synonym(s):

Potassium platinum(II) chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2PtCl4
CAS Number:
Molecular Weight:
415.09
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
powder

grade

for analytical purposes

Assay

99.99% trace metals basis

form

powder

impurities

≤150.0 ppm Trace Metal Analysis

mp

500 °C (lit.)

density

3.38 g/mL at 25 °C (lit.)

SMILES string

[K+].[K+].Cl[Pt--](Cl)(Cl)Cl

InChI

1S/4ClH.2K.Pt/h4*1H;;;/q;;;;2*+1;+2/p-4

InChI key

RVRDLMCWUILSAH-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

Reacts with bis(dithiolates) to form metal-bis(dithiolates) with applications in laser Q-switch materials, liquid crystals, optical CD recording media, bar code materials and superconductivity. Reacts with capping polymers to form shape-control colloidal platinum nanoparticles for catalysis and photocatalysts.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kamel Eid et al.
Nanoscale, 11(24), 11755-11764 (2019-06-12)
The tailored synthesis of carbon nitrides (CNs) is of particular interest in multidisciplinary catalytic applications. However, their fabrication in the form of one-dimensional (1D) nanorods for electrocatalytic carbon monoxide (CO) oxidation is not hitherto reported. Herein, a facile roadmap is
P Koch et al.
Contact dermatitis, 34(4), 253-257 (1996-04-01)
We report a patient with contact stomatitis due to combined sensitization to palladium and platinum. Patch testing showed strong and persistent allergic patch test reactions to palladium chloride (1% pet.), ammonium tetrachloroplatinate (0.25% pet.), and a palladium metal plate. A
Erik Ullerås et al.
Toxicology, 206(2), 245-256 (2004-12-14)
Predictive testing of immunotoxicity associated with chemical compounds is complicated and cannot be accomplished with a single test. As most of the existing tests for immunotoxicity employ experimental animals, there is an increasing need for alternative tests in vitro. We
Gordana N Ostojic et al.
Langmuir : the ACS journal of surfaces and colloids, 24(17), 9784-9789 (2008-07-24)
A method for noncovalent functionalization of DNA-wrapped single-walled carbon nanotubes (SWNTs) using platinum-based DNA cross-linkers is investigated. In particular, cisplatin and potassium tetrachloroplatinate are shown to bind to DNA that encapsulates SWNTs in aqueous solution. The bound platinum salt can
H C Schuppe et al.
International archives of allergy and immunology, 112(2), 125-132 (1997-02-01)
The sensitizing properties of different complex salts of platinum were defined in vivo by means of the popliteal lymph node (PLN) assay in mice. Hexa- and tetrachloroplatinates were confirmed to be highly immunogenic, inducing vigorous primary immune responses in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service