Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Potassium tetrachloroplatinate(II)
Synonym(s):
Potassium platinum(II) chloride
Linear Formula:
K
2
PtCl
4
CAS Number:
10025-99-7
Molecular Weight:
415.09
Products
Brand
Sigma-Aldrich (4)
Formula Weight
Physical Form
Purity
Chemical Composition
Quality Segment
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
206075
Potassium tetrachloroplatinate(II),
98%
Expand
View Pricing
520853
Potassium tetrachloroplatinate(II),
≥99.9% trace metals basis
Expand
View Pricing
323411
Potassium tetrachloroplatinate(II),
99.99% trace metals basis
Expand
View Pricing
925098
Potassium tetrachloroplatinate(II),
Technipur
®
, ≥99.9% trace metals basis
Expand
View Pricing