Skip to Content
Merck
All Photos(2)

Key Documents

252514

Sigma-Aldrich

2-Chloroethyl chloroformate

97%

Synonym(s):

2-Chloroethoxycarbonyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCOOCH2CH2Cl
CAS Number:
Molecular Weight:
142.97
Beilstein:
506639
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

0.23 psi ( 20 °C)

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.446 (lit.)

bp

155-156 °C (lit.)

density

1.385 g/mL at 25 °C (lit.)

functional group

chloro

storage temp.

2-8°C

SMILES string

ClCCOC(Cl)=O

InChI

1S/C3H4Cl2O2/c4-1-2-7-3(5)6/h1-2H2

InChI key

SVDDJQGVOFZBNX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Chloroethyl chloroformate has been used in the synthesis of:
  • (4S)-4-hydroxymethyl-N-[(αR)-α-methylbenzyl]-2-oxazolidinone
  • propargyl dienamide (ll), a precursor of allenic dienamide
  • (±)-7-(2-chloroethyloxycarbonyloxy)-6-(5-chloropyridin-2-yl)-6,7-dihydro-5H-pyrrolo-[3,4-b]-pyrazin-5-one

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

158.0 °F - closed cup

Flash Point(C)

70 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Asymmetric synthesis of the 4-hydroxymethyl-2-oxazolidinone from the serinol derivative and chloroformates.
Sugiyama S, et al.
Tetrahedron Letters, 40(42), 7489-7492 (1999)
Enzymatic resolution of new carbonate intermediates for the synthesis of (< i> S</i>)-(+)-zopiclone.
Solares LF, et al.
Tetrahedron Asymmetry, 13(23), 2577-2582 (2002)
A new approach to pyrrolophenanthridone alkaloids via allene intramolecular cycloaddition: Total synthesis of Hippadine.
Hayakawa K, et al.
Tetrahedron Letters, 28(47), 5895-5898 (1987)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service