Skip to Content
Merck
All Photos(5)

Key Documents

330752

Sigma-Aldrich

Triphosgene

reagent grade, 98%

Synonym(s):

Bis(trichloromethyl) carbonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl3COCOOCCl3
CAS Number:
Molecular Weight:
296.75
Beilstein:
1787583
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

grade

reagent grade

Assay

98%

form

powder

reaction suitability

reaction type: Coupling Reactions

bp

203-206 °C (lit.)

mp

79-83 °C (lit.)

functional group

carbonate
chloro

storage temp.

2-8°C

SMILES string

ClC(Cl)(Cl)OC(=O)OC(Cl)(Cl)Cl

InChI

1S/C3Cl6O3/c4-2(5,6)11-1(10)12-3(7,8)9

InChI key

UCPYLLCMEDAXFR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Triphosgene [Bis(trichloromethyl)carbonate] is a versatile organic reagent used in organic synthesis alternative to phosgene. A catalytic amount of triphosgene is particularly used in chloroformylations, carbonylations, chlorinations, and dehydration reactions.
It is also commonly employed as a coupling agent in the synthesis of carbonyl compounds.

Application

Triphosgene can be employed as a reagent to prepare:
  • Thiocarbonates from thiols and alcohols by one-pot, three-component reaction.
  • Substituted azetidin-2-ones from acids and imines via ketene–imine cycloaddition reaction.
  • Methyl (S)-2-isocyanato-3-phenylpropanoate from L-phenylalanine methyl ester hydrochloride in the presence of sodium bicarbonate.
  • Acyl azides derivatives from various carboxylic acids and sodium azide.
  • Immunosuppressant agent cyclosporin by solid-phase peptide synthesis.
  • Allyl azides from allyl alcohols and sodium azide in one pot method.
  • Esterification coupling reagent di-2-thienyl carbonate, from 2(5H)-thiophenone.
  • 2-Chloronicotinaldehydes via cyclization of the corresponding enamides.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Coordination of chiral amines to coordinatively unsaturated Cp* Ir-amino acid complexes allows determination of enantiomeric purity
Grotjahn DB and Joubran C
Tetrahedron Asymmetry, 6, 745-745 (1995)
Convenient route to thiocarbonates from alcohols, thiols, and triphosgene
Movassagh B and Soleiman-Beigi M
Synthetic Communications, 40(23), 3467-3471 (2010)
A simple one-pot method for the preparation of allyl azides from allyl alcohols using triphosgene: Synthesis of N1-cinnamyl azetidin-2-ones
Jayanthi A, et al.
Synlett, 6, 979-982 (2004)
Aldrichimica Acta, 21, 47-47 (1988)
Triphosgene, a crystalline phosgene substitute
Eckert, Heiner and Forster, Barbara
Angewandte Chemie (International Edition in English), 26(9), 894-895 (1987)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service