Skip to Content
Merck
All Photos(2)

Documents

C41134

Sigma-Aldrich

2-Chloroethyl ether

99%

Synonym(s):

Bis(2-chloroethyl) ether, 2,2′-Dichlorodiethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(ClCH2CH2)2O
CAS Number:
Molecular Weight:
143.01
Beilstein:
605317
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

0.4 mmHg ( 20 °C)

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.456 (lit.)

bp

65-67 °C/15 mmHg (lit.)

mp

−47 °C (lit.)

density

1.22 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

ClCCOCCCl

InChI

1S/C4H8Cl2O/c5-1-3-7-4-2-6/h1-4H2

InChI key

ZNSMNVMLTJELDZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Carc. 2 - Flam. Liq. 3

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

131.0 °F - closed cup

Flash Point(C)

55.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alex Christensen et al.
Water research, 43(16), 3910-3921 (2009-05-12)
The objective of this study was to evaluate the extent of improvement in the biodegradability of persistent organic compounds by pre-oxidation by using Sequential Batch Reactors (SBRs). Dichlorodiethyl ether (DCDE), a non-biodegradable compound, was used as a test chemical. Ozonation
Maria D M Mutuc et al.
Chemosphere, 70(8), 1390-1398 (2007-12-07)
This study examined the feasibility of using surface catalyzed Fenton treatment to remediate soil and groundwater contaminated by the chlorinated ethers, bis(2-chloroethyl) ether (BCEE) and bis(2-chloroethoxy) methane (BCEM). Parameters that affect the contaminant loss rate such as porewater pH, hydrogen
Kevin McClay et al.
Applied and environmental microbiology, 73(21), 6870-6875 (2007-09-18)
Degradation of bis(2-chloroethyl) ether (BCEE) was observed to occur in two bacterial strains. Strain ENV481, a Xanthobacter sp. strain, was isolated by enrichment culturing of samples from a Superfund site located in the northeastern United States. The strain was able
L M Gwinner et al.
Carcinogenesis, 4(11), 1483-1486 (1983-11-01)
The roles of chloroethylene oxide (CEO) and chloroacetaldehyde (CAA) in carcinogenicity of vinyl chloride (VC) have been studied by comparing biological effects of VC exposure with those of 2,2'-dichlorodiethylether (bis(chloroethyl)ether, BCEE) as a metabolic precursor of CAA. Biological end-points investigated
R D Lingg et al.
Archives of environmental contamination and toxicology, 11(2), 173-183 (1982-01-01)
Male rats were given single peroral doses of bis(1-14C-2-chloroethyl)ether ([1-14C]BCEE) (40 mg/kg) and of bis(1-14C-2-chloroisopropyl)ether ([1-14C]BCIE) (90 mg/kg). Excretion of 14CO2 and urinary 14C was followed for 48 hr. The time required to eliminate one half of the dose was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service