Skip to Content
Merck
All Photos(3)

Key Documents

226920

Sigma-Aldrich

2,2-Dimethyl-1,3-propanediamine

99%

Synonym(s):

1,3-Diamino-2,2-dimethylpropane, 2,2-Dimethyltrimethylenediamine, Neopentanediamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NCH2C(CH3)2CH2NH2
CAS Number:
Molecular Weight:
102.18
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

99%

refractive index

n20/D 1.4566 (lit.)

bp

152-154 °C (lit.)

mp

29-31 °C (lit.)

density

0.851 g/mL at 25 °C (lit.)

SMILES string

CC(C)(CN)CN

InChI

1S/C5H14N2/c1-5(2,3-6)4-7/h3-4,6-7H2,1-2H3

InChI key

DDHUNHGZUHZNKB-UHFFFAOYSA-N

Gene Information

General description

2,2-Dimethyl-1,3-propanediamine, commonly referred to as neopentanediamine, is available in solid form with an assay of 99%. It serves as a versatile building block in the synthesis of various polymers. Its two primary amine groups facilitate reactions that lead to the formation of polyamides and polyurethanes, enhancing the mechanical properties and thermal stability of these materials.

Application

2,2-Dimethyl-1,3-propanediamine can be used as a bidentate ligand in the preparation of complexes with aluminum, gallium, and copper. These metal complexes can serve as catalysts in various chemical reactions. It is also utilized as a diamine component in the production of polyamides and polyurethanes. Its two amine groups allow for the formation of strong covalent bonds with diacids or diisocyanates, leading to the development of high-performance polymeric materials with enhanced mechanical properties and thermal stability. It is also used to create hydrogels or other biocompatible materials for applications in tissue engineering and drug delivery systems. Due to its diamine structure, it might function as a chelating agent, useful in treating heavy metal poisoning by binding metal ions. It could potentially be involved in the synthesis of contrast agents used in medical imaging technologies.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

125.6 °F

Flash Point(C)

52 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bhuvanenthiran Mutharani et al.
Ultrasonics sonochemistry, 56, 410-421 (2019-05-19)
To dates, the facile synthesis of inorganic-coated organic polymer composite has received greater attention in the order of research fields including advanced materials and electrochemical analysis owing to the complementary or synergistic effects. In this context, Pr2O3 and Pr2O3 coated
Morteza Montazerozohori et al.
Ultrasonics sonochemistry, 38, 134-144 (2017-06-22)
A bidentate Schiff base ligand and its nano-structured cobalt(II) complex were synthesized under ultrasound irradiation and then their structures were identified by physical and spectral techniques SEM and XRD techniques were used to confirm the nanostructure character of the complex.
Ismail Warad et al.
Ultrasonics sonochemistry, 52, 428-436 (2018-12-24)
Under mode of ultrasonic vibration, the neutral octahedral mononuclear [trans-CuBr2(N ∩ N)2]·3H2O complex with N ∩ N = 2,2-dimethylpropane-1,3-diamine was obtained. The structure of the desired complex was characterized by UV-Vis. spectroscopy, FT-IR, EDX, MS, SEM, TG/DTA and CHN-analysis. The octahedral-structure of the desired Cu(II) complex

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service