Skip to Content
Merck
All Photos(1)

Key Documents

127027

Sigma-Aldrich

N-Methyl-1,3-diaminopropane

98%

Synonym(s):

3-(Methylamino)propylamine, N-Methyl-1,3-propanediamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3NH(CH2)3NH2
CAS Number:
Molecular Weight:
88.15
Beilstein:
878143
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.447 (lit.)

bp

139-141 °C (lit.)

density

0.844 g/mL at 25 °C (lit.)

SMILES string

CNCCCN

InChI

1S/C4H12N2/c1-6-4-2-3-5/h6H,2-5H2,1H3

InChI key

QHJABUZHRJTCAR-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

N-Methyl-1,3-diaminopropane (MeDAP) has been used as a structure directing molecule in the synthesis of two crystalline aluminophosphates. It has also been used in the synthesis of an intermediate of alfuzosin and lapyrium chloride with lipases as catalysts.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1A

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

107.6 °F - DIN 51755 Part 1

Flash Point(C)

42 °C - DIN 51755 Part 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and characterization of two aluminophosphates templated byN-methyl-1, 3-diaminopropane.
Tuel A, et al.
Journal of Solid State Chemistry, 178(3), 782-791 (2005)
Leslie S Hamachi et al.
Chemical science, 10(26), 6539-6552 (2019-08-02)
We report a method to control the composition and microstructure of CdSe1-x S x
Alicia Baldessari
Methods in molecular biology (Clifton, N.J.), 861, 445-456 (2012-03-20)
The application of lipases as catalysts in the synthesis of an intermediate of alfuzosin and lapyrium chloride is described. In the first case, the one-pot procedure to obtain the intermediate involves the treatment of tetrahydrofuroic acid with ethanol in the
Bum-Ho Bin et al.
PloS one, 10(6), e0129273-e0129273 (2015-06-10)
The SLC45A2 gene encodes a Membrane-Associated Transporter Protein (MATP). Mutations of this gene cause oculocutaneous albinism type 4 (OCA4). However, the molecular mechanism of its action in melanogenesis has not been elucidated. Here, we discuss the role of MATP in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service