Skip to Content
Merck
All Photos(1)

Key Documents

33262

Sigma-Aldrich

1,3-Diamino-2-propanol

purum, ≥96.5% (GC)

Synonym(s):

1,3-Diamino-2-hydroxypropane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CH2CH(OH)CH2NH2
CAS Number:
Molecular Weight:
90.12
Beilstein:
741859
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

purum

Quality Level

Assay

≥96.5% (GC)

form

solid

impurities

≤2.0% water

mp

40-44 °C (lit.)

solubility

water: soluble 1 g/10 mL

functional group

amine
hydroxyl

storage temp.

2-8°C

SMILES string

NCC(O)CN

InChI

1S/C3H10N2O/c4-1-3(6)2-5/h3,6H,1-2,4-5H2

InChI key

UYBWIEGTWASWSR-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

1,3-Diamino-2-propanol was used in the synthesis of poly(2-hydroxypropylene imine), poly(2-hydroxypropylene imine ethylene imine) and poly(hydroxypropylene imine propylene imine). It was also used in the synthesis of tetranuclear mixed ligand copper(II) complex of a pyrazole containing Schiff base and a hydroxyhexahydropyrimidylpyrazole group.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sachindranath Pal et al.
Inorganic chemistry, 44(11), 3880-3889 (2005-05-24)
A tetranuclear mixed ligand copper(II) complex of a pyrazole containing Schiff base and a hydroxyhexahydropyrimidylpyrazole and copper(II) and nickel(II) complexes of the Schiff base having N-donor atoms have been investigated. A 2 equiv amount of 5-methyl-3-formylpyrazole (MPA) and 2 equiv
L Alhonen-Hongisto
The Biochemical journal, 190(3), 747-754 (1980-09-15)
1. The mechanism of stimulation of S-adenosylmethionine decarboxylase (EC 4.1.1.50) activity by inhibitors of ornithine decarboxylase (EC 4.1.1.17), namely dl-alpha-difluoromethylornithine, 1,3-diaminopropane and 1,3-diaminopropan-2-ol, was studied in Ehrlich ascites-tumour cells grown in suspension cultures. 2. Difluoromethylornithine and diaminopropane, although decreasing the
Y W Hu et al.
Canadian journal of physiology and pharmacology, 60(12), 1493-1498 (1982-12-01)
The effect of diaminopropanol, an inhibitor of polyamine synthesis, on the metabolic response of liver to insulin was studied in streptozotocin-diabetic rats. Insulin elicited a prompt and very marked increase in ornithine and S-adenosylmethionine decarboxylase activities and in putrescine concentration.
M Mach et al.
The Biochemical journal, 202(1), 153-162 (1982-01-15)
Inhibitors of polyamine synthesis (alpha-methylornithine and 1,3-diaminopropan-2-ol) were used to study the relationship between polyamine synthesis and specific methylations of tRNA in Dictyostelium discoideum during vegetative growth. Polyamine concentrations were found to be 10 mM for putrescine, 1.6 mM for
J M Matés et al.
Biochemical pharmacology, 42(5), 1045-1052 (1991-08-08)
Ornithine decarboxylase (ODC) activity of Ehrlich carcinoma cells was increased more than 36-fold after being maintained for 3.5 hr in vitro in a special chamber which allowed continuous perifusion with 0.5 mM ornithine; if incubated in vitro without perifusion the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service