Skip to Content
Merck
All Photos(3)

Key Documents

D145009

Sigma-Aldrich

3-(Dimethylamino)-1-propylamine

99%

Synonym(s):

N,N-Dimethyl-1,3-diaminopropane, N,N-Dimethyl-1,3-propanediamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2N(CH2)3NH2
CAS Number:
Molecular Weight:
102.18
Beilstein:
605293
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

3.6 (vs air)

vapor pressure

5 mmHg ( 20 °C)

Assay

99%

form

liquid

refractive index

n20/D 1.435 (lit.)

bp

133 °C (lit.)

density

0.812 g/mL at 25 °C (lit.)

SMILES string

CN(C)CCCN

InChI

1S/C5H14N2/c1-7(2)5-3-4-6/h3-6H2,1-2H3

InChI key

IUNMPGNGSSIWFP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3-(Dimethylamino)-1-propylamine is widely used to functionalize various pharmacologically active medicinally important compounds. It is also used in the preparation of dimethylaminopropylamine surfactants that exhibit biological activity against Gram-positive and negative bacteria and fungi.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

89.6 °F - closed cup

Flash Point(C)

32 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Novel 4-aminoquinazoline derivatives induce growth inhibition, cell cycle arrest and apoptosis via PI3Kα inhibition
Fan Y, et al.
Bioorganic & Medicinal Chemistry, 26(8), 1675-1685 (2018)
Synthesis, surface, thermodynamic properties and biological activity of dimethylaminopropylamine surfactants
Shaban SM, et al.
Journal of Industrial and Engineering Chemistry, 20(6), 4194-4201 (2014)
J F Fowler et al.
Contact dermatitis, 37(6), 276-281 (1998-02-10)
Cocamidopropyl betaine (CAPB) is an amphoteric surfactant commonly used in personal care products and surface cleaners. Patch testing with commercially-available CAPB has yielded occasional reactions indicative of allergic contact dermatitis. To determine if subjects with previous positive patch tests would
Ga Young Park et al.
Journal of the American Chemical Society, 134(20), 8513-8524 (2012-05-11)
Certain side-on peroxo-dicopper(II) species with particularly low ν(O-O) (710-730 cm(-1)) have been found in equilibrium with their bis-μ-oxo-dicopper(III) isomer. An issue is whether such side-on peroxo bridges are further activated for O-O cleavage. In a previous study (Liang, H.-C., et
Carrier and inhibitory effects of surfactants on allergic contact reactions to 3-dimethylaminopropylamine.
G Angelini et al.
Contact dermatitis, 39(3), 152-153 (1998-10-15)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service