122017
o-Toluoyl chloride
99%
Synonym(s):
2-Methylbenzoyl chloride
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CH3C6H4COCl
CAS Number:
Molecular Weight:
154.59
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
99%
refractive index
n20/D 1.5549 (lit.)
bp
88-90 °C/12 mmHg (lit.)
density
1.185 g/mL at 25 °C (lit.)
functional group
acyl chloride
SMILES string
Cc1ccccc1C(Cl)=O
InChI
1S/C8H7ClO/c1-6-4-2-3-5-7(6)8(9)10/h2-5H,1H3
InChI key
GPZXFICWCMCQPF-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Flash vacuum pyrolysis of o-toluoyl chloride at 780°C and 0.5 mmHg yields benzocyclobutenone.
Application
- Development of hydrophobic functionalized materials: o-Toluoyl chloride was utilized in the synthesis of novel hydrophobic functionalized UiO-66 materials. The study demonstrated the chemical′s role in enhancing the structural and physicochemical properties of these metal-organic frameworks, indicating potential applications in catalysis and adsorption processes (Narea et al., 2023).
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
170.6 °F - closed cup
Flash Point(C)
77 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Improved synthesis of benzocyclobutenone by flash vacuum pyrolysis.
Suzzarini L, et al.
Tetrahedron Letters, 39(13), 1695-1696 (1998)
Ataf Ali Altaf et al.
Dalton transactions (Cambridge, England : 2003), 47(34), 11769-11781 (2018-08-18)
Twenty-three compounds in two series of ferrocene-based anilides, with the general formula C5H5-Fe-C5H4-C6H4-NH-CO-C6H4-R (where R = H, F, Cl, CH3 and OCH3), have been successfully synthesized. The compounds were characterized by elemental analysis and FTIR, 1H NMR and 13C NMR
Shigeo Hayashi et al.
Journal of enzyme inhibition and medicinal chemistry, 29(6), 846-867 (2014-02-13)
Because of the pivotal role of cyclooxygenase (COX) in the inflammatory processes, non-steroidal anti-inflammatory drugs (NSAIDs) that suppress COX activities have been used clinically for the treatment of inflammatory diseases/syndromes; however, traditional NSAIDs exhibit serious side-effects such as gastrointestinal damage
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service