Skip to Content
Merck
All Photos(3)

Key Documents

111910

Sigma-Aldrich

2-Chlorotoluene

99%

Synonym(s):

1-chloro-2-methylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4Cl
CAS Number:
Molecular Weight:
126.58
Beilstein:
1904175
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39050205
PubChem Substance ID:
NACRES:
NA.22

vapor density

4.38 (vs air)

Quality Level

vapor pressure

10 mmHg ( 43 °C)

Assay

99%

form

liquid

refractive index

n20/D 1.525 (lit.)

bp

157-159 °C (lit.)

mp

−36 °C (lit.)

solubility

H2O: soluble

density

1.083 g/mL at 25 °C (lit.)

functional group

chloro

SMILES string

Cc1ccccc1Cl

InChI

1S/C7H7Cl/c1-6-4-2-3-5-7(6)8/h2-5H,1H3

InChI key

IBSQPLPBRSHTTG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Chlorotoluene can be mineralized completely by Rhodococcus, strain OCT 10 DSM 45596(T). It is used in the preparation of 3-chloro-4-methylcatechol and 4-chloro-3-methylcatechol. The oxidation of 2-chlorotoluene is very well catalyzed by the enzymes: toluene dioxygenase of P. putida F1 and chlorobenzene dioxygenase from Burkholderia, strain PS12.

Application

2-Chlorotoluene can be used as a precursor to synthesize aryl coupling products via palladium-catalyzed Negishi cross-coupling. It can also be used in the Buchwald-Hartwig amination to synthesize arylamines.

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Acute 1 - Aquatic Chronic 2 - Flam. Liq. 3 - Repr. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

109.4 °F - closed cup

Flash Point(C)

43 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

NIXANTPHOS: A Highly Active Ligand for Palladium Catalyzed Buchwald-Hartwig Amination of Unactivated Aryl Chlorides.
Mao J, et al.
Dalton Transactions, (26) (2018)
A Lehning et al.
Applied and environmental microbiology, 63(5), 1974-1979 (1997-05-01)
The degradation of toluene by Pseudomonas putida F1 and of chlorobenzenes by Burkholderia sp. strain PS12 is initiated by incorporation of dioxygen into the aromatic nucleus to form cis-dihydrodihydroxybenzenes. Toluene-grown cells of P. putida F1 and 3-chlorobenzoate-grown cells of Burkholderia
Flexible Steric Bulky Bis (Imino) acenaphthene (BIAN)-Supported N-Heterocyclic Carbene Palladium Precatalysts: Catalytic Application in Buchwald?Hartwig Amination in Air.
Lan X B, et al.
The Journal of Organic Chemistry, 82(6), 2914-2925 (2017)
The first general method for palladium-catalyzed Negishi cross-coupling of aryl and vinyl chlorides: use of commercially available Pd (P (t-Bu) 3) 2 as a catalyst
Dai C, et al.
Journal of the American Chemical Society, 123(12), 2719-2724 (2001)
Katrin Pollmann et al.
Microbiology (Reading, England), 149(Pt 4), 903-913 (2003-04-11)
The tetrachlorobenzene dioxygenase (TecA) of Ralstonia sp. PS12 carries out the first step in the aerobic biodegradation of chlorinated toluenes. Besides dioxygenation of the aromatic ring of 4-chloro-, 2,4-, 2,5- and 3,4-dichlorotoluene as the main reaction, it also catalyses mono-oxygenation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service