Skip to Content
Merck
All Photos(2)

Key Documents

119946

Sigma-Aldrich

4-Fluorobenzoyl chloride

98%

Synonym(s):

4-Fluorobenzoic acid chloride, p-Fluorobenzoyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H4COCl
CAS Number:
Molecular Weight:
158.56
Beilstein:
386215
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

refractive index

n20/D 1.532 (lit.)

bp

82 °C/20 mmHg (lit.)

mp

10-12 °C (lit.)

density

1.342 g/mL at 25 °C (lit.)

functional group

acyl chloride
fluoro

SMILES string

Fc1ccc(cc1)C(Cl)=O

InChI

1S/C7H4ClFO/c8-7(10)5-1-3-6(9)4-2-5/h1-4H

InChI key

CZKLEJHVLCMVQR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Fluorobenzoyl chloride undergoes Friedel-Crafts acylation reaction with 2,6-dimethylnaphthalene to give 1,5-bis(4-fluorobenzoyl)-2,6-dimethylnaphthalene.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

179.6 °F - closed cup

Flash Point(C)

82 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Novel electroactive aromatic polyamide with oligoanilines and azo groups in the backbone: synthesis, characterization and dielectric properties.
He L, et al.
Journal of Polymer Research, 19(1), 1-9 (2012)
New multiblock copolymers of sulfonated poly (4'-phenyl-2, 5-benzophenone) and poly (arylene ether sulfone) for proton exchange membranes. II.
Ghassemi H, et al.
Polymer, 45(17), 5855-5862 (2004)
Synthesis of a novel naphthalene-based poly (arylene ether-ketone) by polycondensation of 1, 5-bis (4-fluorobenzoyl)-2, 6-dimethylnaphthalene with bisphenol A.
Ohno M, et al.
Journal of Polymer Science Part A: Polymer Chemistry, 33(15), 2647-2655 (1995)
Synthesis of [18F] GBR 12909, a dopamine reuptake inhibitor.
Haka MS and Kilbourn MR.
Journal of Labelled Compounds & Radiopharmaceuticals, 28(7), 793-799 (1990)
Solid-phase Friedel-Crafts acylation on polystyrene resins-synthesis of antiepiletic 1-aryl-3, 5-dihydro-4H-2, 3-benzodiazepin-4-ones.
Bevacqua F, et al.
Tetrahedron Letters, 42(43), 7683-7685 (2001)

Articles

Friedel-Crafts acylation with Lewis acid catalysts forms monoacylated products via electrophilic aromatic substitution of arenes.

Friedel-Crafts acylation with Lewis acid catalysts forms monoacylated products via electrophilic aromatic substitution of arenes.

Friedel-Crafts acylation with Lewis acid catalysts forms monoacylated products via electrophilic aromatic substitution of arenes.

Friedel-Crafts acylation with Lewis acid catalysts forms monoacylated products via electrophilic aromatic substitution of arenes.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service