Skip to Content
Merck
All Photos(3)

Documents

P16753

Sigma-Aldrich

Phenylacetyl chloride

98%

Synonym(s):

α-Toluyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2COCl
CAS Number:
Molecular Weight:
154.59
Beilstein:
742254
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.5325 (lit.)

bp

94-95 °C/12 mmHg (lit.)

density

1.169 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

ClC(=O)Cc1ccccc1

InChI

1S/C8H7ClO/c9-8(10)6-7-4-2-1-3-5-7/h1-5H,6H2

InChI key

VMZCDNSFRSVYKQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Phenylacetyl chloride is used as a key precursor in the total synthesis of vialinin B. It is employed as a linker to prepare dendrimers and also used in the synthesis of various conjugated aromatic small molecules.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

215.6 °F

Flash Point(C)

102 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Xiao-Yan Zhang et al.
Journal of agricultural and food chemistry, 66(34), 8976-8982 (2018-08-11)
Five new resorcylic acid lactones (RALs) hispidulactones A-E (1, 4, 5, 8, and 9), a new natural product (2), and four known ones (3, 6, 7, and 10) with different ring systems were isolated from the desert plant endophytic fungus
A Light-Harvesting Array Composed of Porphyrins and Rigid Backbones.
Kozaki M, et al.
Organic Letters, 10(20), 4477-4480 (2008)
Expeditious synthesis of vialinin B, an extremely potent inhibitor of TNF-α production.
Ye Y Q, et al.
Organic Letters, 11(21), 5074-5077 (2009)
Solution-processable α, ω-distyryl oligothiophene semiconductors with enhanced environmental stability.
Mauldin C E, et al.
Chemistry of Materials, 21(9), 1927-1938 (2009)
New electron-accepting π-conjugated polyquinoxalines with fluorene unit: synthesis and light-emitting properties.
Jung S H, et al.
Polym. Bull., 50(4), 251-258 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service