Merck
  • Shp2 regulates chlorogenic acid-induced proliferation and adipogenic differentiation of bone marrow-derived mesenchymal stem cells in adipogenesis.
Shp2 regulates chlorogenic acid-induced proliferation and adipogenic differentiation of bone marrow-derived mesenchymal stem cells in adipogenesis.
Molecular medicine reports (2015-01-31)
Rong-Ping Zhou, Ming-Tao Deng, Lan-Ying Chen, Ning Fang, Chuan Du, Lin-Pan Chen, Ye-Qing Zou, Jiang-Hua Dai, Mei-Lan Zhu, Wei Wang, Si-Jian Lin, Rong-Hua Liu, Jun Luo
ABSTRACT
Chlorogenic acid (CGA) exhibits various biological properties, including the inhibition of oxidation, obesity, apoptosis and tumorigenesis. CGA is also able to promote cell survival and proliferation. The aim of the present study was to determine the effects and underlying molecular mechanisms of CGA on the adipogenesis of bone marrow‑derived mesenchymal stem cells (BMSCs). Treatment with CGA had a marginal effect on cell proliferation, by promoting the expression levels of phosphorylated Akt and cyclin D1. Furthermore, treatment with CGA also upregulated the phosphorylation of extracellular signal‑regulated kinase (Erk) and inhibited the adipocyte differentiation of BMSCs by inhibiting the expression of peroxisome proliferator‑activated receptor (PPAR)γ and CCAAT/enhancer binding protein (C/EBP)α. However, knockdown of the expression of Shp2 attenuated CGA‑induced proliferation and inhibited the phosphorylation of Akt and expression of cyclin D1. Furthermore, CGA treatment upregulated Erk phosphorylation and decreased the expression levels of PPARγ and CEBPα, which was inhibited by treatment with the Shp2 PTPase activity inhibitor, NSC‑87877. The results of the present study suggested that CGA‑induced Akt and Erk pathways regulate proliferation and differentiation and that Shp2 is important in the proliferation and differentiation of BMSCs.
Sigma-Aldrich
Sodium dodecyl sulfate, SAJ special grade, ≥97.0%
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Chlorogenic acid, ≥95% (titration)
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 7, ≥98%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
5-O-p-Coumaroylquinic acid, ≥95%
Sigma-Aldrich
4-O-p-Coumaroylquinic acid
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
MISSION® esiRNA, targeting human PTPN11
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Ptpn11
SAFC
Sodium deoxycholate
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Fluorescein, for fluorescence, free acid
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Sodium deoxycholate, ≥97% (titration)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
3-O-p-Coumaroylquinic acid, ≥95%
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)