Skip to Content
Merck
All Photos(1)

Key Documents

28-3260

Sigma-Aldrich

Sodium dodecyl sulfate

SAJ special grade, ≥97.0%

Synonym(s):

Dodecyl sodium sulfate, Dodecyl sulfate sodium salt, Lauryl sulfate sodium salt, SDS, Sodium lauryl sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)11OSO3Na
CAS Number:
Molecular Weight:
288.38
Beilstein:
3599286
EC Number:
MDL number:
UNSPSC Code:
12161902
PubChem Substance ID:

grade

SAJ special grade

vapor pressure

≤0 hPa ( 20 °C)

description

anionic detergent

Assay

≥97.0%

form

rod

mol wt

288.38 g/mol

availability

available only in Japan

pH

9.1 (10 g/L)

mp

204-207 °C (lit.)

SMILES string

[Na+].CCCCCCCCCCCCOS([O-])(=O)=O

InChI

1S/C12H26O4S.Na/c1-2-3-4-5-6-7-8-9-10-11-12-16-17(13,14)15;/h2-12H2,1H3,(H,13,14,15);/q;+1/p-1

InChI key

DBMJMQXJHONAFJ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Anionic detergent

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

338.0 °F

Flash Point(C)

170 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shane Stegeman et al.
Endocrine-related cancer, 22(2), 265-276 (2015-02-12)
The oncogene MDM4, also known as MDMX or HDMX, contributes to cancer susceptibility and progression through its capacity to negatively regulate a range of genes with tumour-suppressive functions. As part of a recent genome-wide association study it was determined that
Kazuya Ishikawa et al.
Nature communications, 5, 5430-5430 (2014-11-13)
Pathogen effector proteins are delivered to host cells to suppress plant immunity. However, the mechanisms by which effector proteins function are largely unknown. Here we show that expression of XopP(Xoo), an effector of rice pathogen Xanthomonas oryzae pv. oryzae, in
S Bavamian et al.
Molecular psychiatry, 20(5), 573-584 (2015-01-28)
Bipolar disorder (BD) is a heritable neuropsychiatric disorder with largely unknown pathogenesis. Given their prominent role in brain function and disease, we hypothesized that microRNAs (miRNAs) might be of importance for BD. Here we show that levels of miR-34a, which
Hannah M Kaneb et al.
Human molecular genetics, 24(5), 1363-1373 (2014-10-26)
Amyotrophic lateral sclerosis (ALS) is a fatal neurodegenerative disorder characterized by the selective death of motor neurons. Causative mutations in the global RNA-processing proteins TDP-43 and FUS among others, as well as their aggregation in ALS patients, have identified defects
Tadayoshi Hayata et al.
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research, 30(2), 318-329 (2014-08-27)
Transforming growth factor (TGF)-β signaling plays critical roles during skeletal development and its excessive signaling causes genetic diseases of connective tissues including Marfan syndrome and acromelic dysplasia. However, the mechanisms underlying prevention of excessive TGF-β signaling in skeletogenesis remain unclear.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service