Skip to Content
Merck
All Photos(2)

Key Documents

71736

Sigma-Aldrich

Sodium dodecyl sulfate solution

BioUltra, for molecular biology, 10% in H2O

Synonym(s):

Lauryl sulfate sodium salt, SDS, Sodium lauryl sulfate solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)11OSO3Na
CAS Number:
Molecular Weight:
288.38
Beilstein:
3599286
MDL number:
UNSPSC Code:
12161902
eCl@ss:
39093306
PubChem Substance ID:
NACRES:
NA.26
Pricing and availability is not currently available.

grade

for molecular biology

Quality Level

description

anionic

product line

BioUltra

form

liquid

mol wt

288.38 g/mol

concentration

10% in H2O

technique(s)

HPLC: suitable
protein quantification: suitable

impurities

DNases, none detected
RNases, none detected
insoluble matter, passes filter test
phosphatases, none detected
proteases, none detected

pH

5.0-6.5

density

1.01 g/mL at 20 °C

anion traces

chloride (Cl-): ≤50 mg/kg
phosphate (PO43-): ≤1 mg/kg

cation traces

Al: ≤1 mg/kg
As: ≤0.1 mg/kg
Ba: ≤1 mg/kg
Bi: ≤1 mg/kg
Ca: ≤5 mg/kg
Cd: ≤1 mg/kg
Co: ≤1 mg/kg
Cr: ≤1 mg/kg
Cu: ≤1 mg/kg
Fe: ≤1 mg/kg
K: ≤20 mg/kg
Li: ≤1 mg/kg
Mg: ≤1 mg/kg
Mn: ≤1 mg/kg
Mo: ≤1 mg/kg
Ni: ≤1 mg/kg
Pb: ≤1 mg/kg
Sr: ≤1 mg/kg
Zn: ≤1 mg/kg

λ

neat

UV absorption

λ: 260 nm Amax: 0.3
λ: 280 nm Amax: 0.2

SMILES string

[Na+].CCCCCCCCCCCCOS([O-])(=O)=O

InChI

1S/C12H26O4S.Na/c1-2-3-4-5-6-7-8-9-10-11-12-16-17(13,14)15;/h2-12H2,1H3,(H,13,14,15);/q;+1/p-1

InChI key

DBMJMQXJHONAFJ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Sodium dodecyl sulfate solution has been used:
  • in the preparation of 2x extraction buffer[1]
  • as a component in elution buffer for immunoprecipitation[2]
  • in SDS–polyacrylamide gel electrophoresis (PAGE) analysis[3][4]

Biochem/physiol Actions

Sodium dodecyl sulfate (SDS)/lauryl sulfate, is an ionic detergent has the ability to disrupt biological membranes. It can block ribonuclease (RNase) and deoxyribonuclease (DNase) activity. SDS is one of the main constituents of several reagents, that are used to purify nucleic acids.[5] It is also considered as a permeability enhancer in oral formulations.[6]

Other Notes

In large-scale phenol extraction of RNA - promotes the dissociation of protein from nucleic acids when extracting from biological material[7]

also commonly purchased with this product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Porous nucleating agents for protein crystallization
Khurshid S, et al.
Nature Protocols, 9(7), 1621-1621 (2014)
D.M. Wallace
Methods in Enzymology, 125, 39-39 (1987)
Dictyostelium cultivation, transfection, microscopy and fractionation
Hirst J, et al.
Bio-protocol, 5(11) (2015)
Construction of FWPV Chimaeric MVA
Buttigieg K R and Skinner M A
Bio-protocol, 5(1) (2015)
Resilient Ribonucleases
RNA Methodologies (2010)

Protocols

TRI Reagent enables simultaneous DNA, RNA, and protein isolation with sample prep guidelines and troubleshooting.

TRI Reagent enables simultaneous DNA, RNA, and protein isolation with sample prep guidelines and troubleshooting.

TRI Reagent enables simultaneous DNA, RNA, and protein isolation with sample prep guidelines and troubleshooting.

TRI Reagent enables simultaneous DNA, RNA, and protein isolation with sample prep guidelines and troubleshooting.

Questions

1–3 of 3 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. I WOULD LIKE TO REQUEST THE QUALITY CONFIRMATION OF %CONCENTRATION, SINCE CONCENTRATION IS 10% IN WATER, THERE'S NO INFORMATION THAT WHAT'S % TYPE [%W/V OR %V/V] REF.PRODUCT CODE: 71736

    1 answer
    1. This product is 10% (w/w) solution.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service