Skip to Content
Merck
All Photos(2)

Key Documents

F3651

Sigma-Aldrich

Fluorescein 5(6)-isothiocyanate

≥90% purity (HPLC), powder

Synonym(s):

FITC, Fluorescein isothiocyanate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H11NO5S
CAS Number:
Molecular Weight:
389.38
Beilstein:
1407295
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

product name

Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)

Assay

≥90% (HPLC)

form

powder

color

yellow to orange

solubility

acetone: 1 mg/mL, clear, light yellow to yellow

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-8°C

SMILES string

Oc1ccc2c(Oc3cc(O)ccc3C24OC(=O)c5cc(ccc45)N=C=S)c1

InChI

1S/C21H11NO5S/c23-12-2-5-16-18(8-12)26-19-9-13(24)3-6-17(19)21(16)15-4-1-11(22-10-28)7-14(15)20(25)27-21/h1-9,23-24H

InChI key

MHMNJMPURVTYEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Fluorescein isothiocyanate (FITC) is widely used to attach a fluorescent label to proteins via the amine group. The isothiocyanate group reacts with amino terminal and primary amines in proteins. It has been used for the labeling of proteins including antibodies and lectins.

Application

Fluorescein 5(6)-isothiocyanate has been used:
  • for fluorescence labeling of bacteria
  • as a standard for calibration of fluorometer
  • for the labeling of exosomes
For fluorescent labeling of proteins (e.g. immunofluorescence); H Protein labeling for gel chromatography

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Characterization of proteolytic enzyme activities in macroalgae.
Perez-Llorens JL
European Journal of Phycology, 38, 55-64 (2003)
Effects of supplemental 25-hydroxycholecalciferol on growth performance, small intestinal morphology, and immune response of broiler chickens.
Chou SH
Poultry Science, 88, 2333-2333 (2009)
Thymic exosomes promote the final maturation of thymocytes.
Lundberg V
Scientific Reports, 6, 36479-36479 (2016)
Hermanson GT
Bioconjugate Techniques (2013)
Handbook of Affinity Chromatography (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service