Skip to Content
Merck
All Photos(1)

Key Documents

935573

Sigma-Aldrich

5-O-p-Coumaroylquinic acid

≥95%

Synonym(s):

(1R,3R,4S,5R)-1,3,4-Trihydroxy-5-[[3-(4-hydroxyphenyl)-1-oxo-2-propen-1-yl]oxy]cyclohexanecarboxylicacid,, Cyclohexanecarboxylic acid, Cyclohexanecarboxylic acid, 1,3,4-trihydroxy-5-[[3-(4-hydroxyphenyl)-1-oxo-2-propenyl]oxy]-, (1R,3R,4S,5R)-, Cyclohexanecarboxylicacid,1,3,4-trihydroxy-5-[[3-(4-hydroxyphenyl)-1-oxo-2-propen-1-yl]oxy]-,(1R,3R,4S,5R)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H18O8
CAS Number:
Molecular Weight:
338.31
UNSPSC Code:
12352106
NACRES:
NA.22

Quality Level

Assay

≥95%

form

powder or crystals

color

white to off-white

bp

613.2 °C

mp

192-199 °C

density

1.55 g/cm3

storage temp.

2-8°C

SMILES string

O=C(OC1CC(O)(C(=O)O)CC(O)C1O)C=CC2=CC=C(O)C=C2

InChI

InChI=1S/C16H18O8/c17-10-4-1-9(2-5-10)3-6-13(19)24-12-8-16(23,15(21)22)7-11(18)14(12)20/h1-6,11-12,14,17-18,20,23H,7-8H2,(H,21,22)/t11-,12-,14+,16-/m1/s1

InChI key

BMRSEYFENKXDIS-UHFFFAOYSA-N

Application

5-O-p-Coumaroylquinic acid has garnered attention in pharmaceutical and nutraceutical industries due to its potential therapeutic applications.

Features and Benefits

5-O-p-Coumaroylquinic acid is a natural phenolic compound that belongs to the class of hydroxycinnamic acid derivatives. It is structurally similar to 4-p-Coumaroylquinic acid, with an additional coumaric acid moiety attached to the 5-position of the quinic acid backbone via an ester linkage. This compound is commonly found in various plant sources and shares similar biological activities with 4-p-Coumaroylquinic acid, including antioxidant, anti-inflammatory, and antimicrobial properties.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Holim Jang et al.
Journal of agricultural and food chemistry, 62(1), 182-191 (2013-12-04)
This study explored whether chlorogenic acid (CGA) and coffee have protective effects against retinal degeneration. Under hypoxic conditions, the viability of transformed retinal ganglion (RGC-5) cells was significantly reduced by treatment with the nitric oxide (NO) donor S-nitroso-N-acetylpenicillamine (SNAP). However
Shengxi Meng et al.
Evidence-based complementary and alternative medicine : eCAM, 2013, 801457-801457 (2013-09-26)
Intracellular glucose and lipid metabolic homeostasis is vital for maintaining basic life activities of a cell or an organism. Glucose and lipid metabolic disorders are closely related with the occurrence and progression of diabetes, obesity, hepatic steatosis, cardiovascular disease, and
M R Olthof et al.
The Journal of nutrition, 131(1), 66-71 (2001-02-24)
Chlorogenic acid, an ester of caffeic acid and quinic acid, is a major phenolic compound in coffee; daily intake in coffee drinkers is 0.5-1 g. Chlorogenic acid and caffeic acid are antioxidants in vitro and might therefore contribute to the
Zekun Liu et al.
Scientific reports, 6, 19095-19095 (2016-01-13)
The Influenza A virus is a great threat for human health, while various subtypes of the virus made it difficult to develop drugs. With the development of state-of-art computational chemistry, computational molecular docking could serve as a virtual screen of
Rong-Ping Zhou et al.
Molecular medicine reports, 11(6), 4489-4495 (2015-01-31)
Chlorogenic acid (CGA) exhibits various biological properties, including the inhibition of oxidation, obesity, apoptosis and tumorigenesis. CGA is also able to promote cell survival and proliferation. The aim of the present study was to determine the effects and underlying molecular

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service