Merck
  • Influence of a Neighboring Charged Group on Hydrophobic Hydration Shell Structure.
Influence of a Neighboring Charged Group on Hydrophobic Hydration Shell Structure.
The journal of physical chemistry. B (2014-11-22)
Joel G Davis, Samual R Zukowski, Blake M Rankin, Dor Ben-Amotz
ABSTRACT
Raman multivariate curve resolution (Raman-MCR), as well as quantum and classical calculations, are used to probe water structural changes in the hydration shells of carboxylic acids and tetraalkyl ammonium ions with various aliphatic chain lengths. The results reveal that water molecules in the hydration shell around the hydrophobic chains undergo a temperature and chain length dependent structural transformation resembling that previously observed in aqueous solutions of n-alcohols. Deprotonation of the carboxylic acid headgroup (at pH ∼ 7) is found to suppress the onset of the hydration-shell structural transformation around the nearest aliphatic methylene group. Tetraalkyl ammonium cations are found to more strongly suppress the water structural transformation, perhaps reflecting the greater intramolecular charge delocalization and suppression of dangling OH defects in water's tetrahedral H-bond network. The observed coupling between ionic and hydrophobic groups, as well as the associated charge asymmetry, may influence the hydrophobicity of proteins and other materials.
Sigma-Aldrich
1-Butanol, suitable for HPLC
Sigma-Aldrich
Octanoic acid, ≥96.0%
Sigma-Aldrich
1-Butanol, JIS special grade, ≥99.0%
Sigma-Aldrich
1-Butanol, SAJ first grade, ≥99.0%
Sigma-Aldrich
Sodium dodecanoate, 99-100%
Supelco
1-Butanol, suitable for HPLC, 99.8%
Sigma-Aldrich
Hexanoic acid, ≥99%
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Octanoic acid, ≥99%
Sigma-Aldrich
Octanoic acid, ≥98%
Sigma-Aldrich
Octanoic acid, ≥98%, FG
Sigma-Aldrich
Octanoic acid, natural, ≥98%, FG
Sigma-Aldrich
1-Propanol, ≥99%, FG
Sigma-Aldrich
1-Propanol, natural, ≥98%, FG
Sigma-Aldrich
Hexanoic acid, ≥98%, FCC, FG
Sigma-Aldrich
Sodium bromide, BioUltra, ≥99.0% (AT)
Sigma-Aldrich
Sodium bromide, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Sodium bromide, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide solution, 1 M
Sigma-Aldrich
Sodium hydroxide solution, 0.05 M
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide solution, 0.1 M
Sigma-Aldrich
Sodium hydroxide, SAJ first grade, ≥95.0%
Sigma-Aldrich
Sodium hydroxide solution, 0.01 M
Sigma-Aldrich
Sodium hydroxide solution, 4 M
Sigma-Aldrich
Sodium hydroxide solution, 6 M
Sigma-Aldrich
Sodium hydroxide, JIS special grade, ≥96.0%
Sigma-Aldrich
1-Propanol, JIS special grade
Sigma-Aldrich
Sodium hydroxide solution, 7 M