Skip to Content
Merck
All Photos(1)

Key Documents

71687

Sigma-Aldrich

Sodium hydroxide

BioUltra, for luminescence, ≥98.0% (T), pellets

Synonym(s):

Caustic Lye, Caustic Soda, Soda Lye, Sodium Hydrate, ‘Caustic soda’

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaOH
CAS Number:
Molecular Weight:
40.00
EC Number:
MDL number:
UNSPSC Code:
12352316
PubChem Substance ID:
NACRES:
NA.32
Pricing and availability is not currently available.

grade

for luminescence

Quality Level

vapor density

>1 (vs air)

vapor pressure

<18 mmHg ( 20 °C)
3 mmHg ( 37 °C)

product line

BioUltra

Assay

≥98.0% (T)

form

pellets

technique(s)

cell culture | mammalian: suitable

impurities

insoluble matter, passes filter test
≤0.0003% total nitrogen (N)
≤0.0005% silicon dioxide (SiO2)
≤1.0% sodium carbonate (Na2CO3)

mp

318 °C (lit.)

solubility

H2O: 1 M at 20 °C, clear, colorless

density

2.13 g/cm3

anion traces

chloride (Cl-): ≤70 mg/kg
phosphate (PO43-): ≤5 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Ag: ≤5 mg/kg
Al: ≤5 mg/kg
As: ≤0.1 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
Hg: ≤0.1 mg/kg
K: ≤500 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

SMILES string

[OH-].[Na+]

λ

1 M in H2O

UV absorption

λ: 260 nm Amax: 0.015
λ: 280 nm Amax: 0.01

suitability

in accordance for luminescence

InChI

1S/Na.H2O/h;1H2/q+1;/p-1

InChI key

HEMHJVSKTPXQMS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium Hydroxide, also known as caustic soda or lye, is a white crystalline odorless solid which is corrosive and absorbs moisture from the air. When dissolved in water, Sodium Hydroxide generates heat to ignite combustible materials. In the form of pellets, Sodium Hydroxide, when dissolved, forms a colorless liquid, denser than water.

Application

Sodium Hydroxide is generally used in the solid or 50% solution form. It is suitable for use at various stages of soaps, paper, rayon, and petroleum product manufacturing. It is also used in carbohydrate research using FTIR analysis.[1] Sodium Hydroxide is used as a pre-treatment reagent to study enzymatic hydrolysis.[2]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ziyu Wang et al.
Bioresource technology, 101(10), 3583-3585 (2010-01-22)
Coastal Bermuda grass was pretreated with NaOH at concentrations from 0.5% to 3% (w/v) for a residence time from 15 to 90min at 121 degrees C. The pretreatments were evaluated based on total lignin removal and production of total reducing
Sang Youn Oh et al.
Carbohydrate research, 340(3), 417-428 (2005-02-01)
Cellulose samples treated with sodium hydroxide (NaOH) and carbon dioxide in dimethylacetamide (DMAc) were analyzed by FTIR spectroscopy. Absorbance of hydrogen-bonded OH stretching was considerably decreased by the treatment of NaOH and carbon dioxide. The relative absorbance ratio (A(4000-2995)/A(993)) represented
Konstantinos A Polyzos et al.
Cardiovascular research, 106(2), 295-302 (2015-03-10)
Atherosclerosis is a chronic inflammatory disease that is initiated by the retention and accumulation of low-density lipoprotein in the artery, leading to maladaptive response of cells from the immune system and vessel wall. Strong evidence implicates indoleamine 2,3-dioxygenase (IDO), the
C Berger et al.
Human reproduction (Oxford, England), 30(4), 800-811 (2015-03-06)
Does ulipristal acetate (UPA) used for emergency contraception (EC) interfere with the human embryo implantation process? UPA, at the dosage used for EC, does not affect human embryo implantation process, in vitro. A single pre-ovulatory dose of UPA (30 mg)
Jesper Hjortnaes et al.
Advanced healthcare materials, 4(1), 121-130 (2014-06-25)
Three dimensional (3D) hydrogel platforms are powerful tools, providing controllable, physiologically relevant microenvironments that could aid in understanding how various environmental factors direct valvular interstitial cell (VIC) phenotype. Continuous activation of VICs and their transformation from quiescent fibroblast to activated

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service